Skip to Content
MilliporeSigma
All Photos(1)

Documents

63325

Millipore

Lactic acid solution

10%, suitable for microbiology, sterile filtered

Synonym(s):

Lactic acid 10% Supplement

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CO2H
CAS Number:
Molecular Weight:
90.08
Beilstein:
5238667
MDL number:
UNSPSC Code:
41171614
NACRES:
NA.71

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10%

application(s)

microbiology

storage temp.

2-8°C

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lactic Acid solution 10% recommended for the adjustment of acidic pH.

Components

Composition (10mL per vial)
Lactic acid 1g
Distilled water 10mL

Other Notes

Contains varying amounts of intermolecular esterification products.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service