Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LSKMPCR

Millipore

MultiScreen-PCRµ96 Filter Plate

Purification of PCR products

Synonym(s):

Montage PCR Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104923
eCl@ss:
32039006
NACRES:
NB.15

material

flat bottom wells
polystyrene

Quality Level

sterility

non-sterile

feature

lid: no

manufacturer/tradename

MultiScreen®

parameter

50-250 μL sample volume (per well)

technique(s)

activity assay: suitable (protein kinase/phosphatase)
bead-based assay: suitable
ligand binding assay: suitable
receptor binding assay: suitable

plate size

96 wells

well maximum volume

175 μL

working volume

100-150 μL

pore size

pore sizeproprietary

binding type

low binding surface

shipped in

ambient

General description

MultiScreen-PCRμ96 filter plate is a small volume, 96-well device for the purification of polymerase chain reaction (PCR) products. The PCRμ96 plate enables purification of both small and large volume PCR reactions (20–100 μL) with the recovery of purified products in as little as 20 μL of solution. The vacuum-based, size exclusion separation effectively removes contaminating salts, unincorporated dNTPs, and primers from PCR reactions in less than 15 minutes. The purified DNA is suitable for downstream applications such as sequencing, genotyping, or micro arraying. Additionally, the automation-friendly design of the MultiScreen PCRμ96 plate makes it ideal for high throughput applications.

Application

MultiScreen-PCRμ96 filter plate has been used in natural product screening, total drug analysis, protein precipitation, bead cleavage, solid-phase synthesis, and solubility assay.It has also been used in the purification of polymerase chain reaction (PCR) products.

Features and Benefits

  • Offers fast, automatable solutions for high-throughput PCR purification.
  • Provides high purity and high recovery.

Legal Information

MULTISCREEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian Stewart et al.
Toxicon : official journal of the International Society on Toxinology, 56(5), 804-812 (2009-08-04)
A referee analysis method for the detection and quantification of Pacific ciguatoxins in fish flesh has recently been established by the public health analytical laboratory for the State of Queensland, Australia. Fifty-six fish samples were analysed, which included 10 fillets
Tristan Barbeyron et al.
International journal of systematic and evolutionary microbiology, 61(Pt 8), 1899-1905 (2010-09-14)
A rod shaped, Gram-stain-negative, chemo-organotrophic, heterotrophic, strictly aerobic, non-gliding bacterium, designated strain PLR(T), was isolated from faeces of the mollusc Aplysia punctata (Mollusca, Gastropoda) that had been fed with green algae belonging to the genus Ulva. The novel strain was
Sarah A Wexler et al.
British journal of haematology, 121(2), 368-374 (2003-04-16)
In postnatal life, mesenchymal stem cells (MSC) self-replicate, proliferate and differentiate into mesenchymal tissues, including bone, fat, tendon, muscle and bone marrow (BM) stroma. Possible clinical applications for MSC in stem cell transplantation have been proposed. We have evaluated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service