Skip to Content
MilliporeSigma
All Photos(2)

Documents

IX70XXPKT1

Milli-Q

Milli-Q® IX Purification Cartridge Kit

This product is replaced with IX70XXPKIT. Search to Explore!

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
NACRES:
NB.85

Quality Level

composition

Soda lime (for CO2 removal)

packaging

pkg of 1 kit

manufacturer/tradename

Milli-Q® IX

compatibility

for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015

Looking for similar products? Visit Product Comparison Guide

General description

This water purification cartridge kit includes the following:
  • IPAK Gard® 10-15 Pretreatment Pack efficiently removes colloids, particles, free chlorine, and organics for reverse osmosis (RO) membrane protection and improved system performance.
  • Tank Vent filter, redesigned for seamless integration, provides improved protection against airborne contaminants.

Application

For optimal protection of the RO membrane and advanced protection against airborne contaminants.

Features and Benefits

IPAK Gard® 10-15 Pretreatment Pack contains:
  • Activated carbon to protect against free chlorine and organics.
  • Depth filter to protect against particulates and colloids.
  • 63% of the parts suppliers for IPAK Gard® cartridge included in this kit are ISO 14001 environmental management system-certified.

Tank Vent Filter contains:
  • Activated carbon to remove volatile organics.
  • Soda lime to remove CO2.
  • Membrane filter to prevent particulates and bacteria from entering the tank.

Legal Information

IPAK GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service