Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

INTELLIFLEX-DRSE

Millipore

xMAP® INTELLIFLEX DRSE

multiplex up to 500 analytes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.84

product line

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric

shipped in

ambient

General description

The xMAP® INTELLIFLEX DRSE System is the latest addition to the Luminex family and has advanced features like an embedded PC with touchscreen and automated maintenance functions. The design reduces the need for optical alignment and is smaller and lighter than other Luminex flow cytometry-based readers. The system runs both 96 and 384 well plates.

Application

The xMAP® INTELLIFLEX DRSE System makes it possible to multiplex up to 500 analytes through the combination of differentially dyed fluorescent microsphere sets & an innovative instrument design. The system is compatible with all the existing MILLIPLEX® Multiplex Assays

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

WEEE Directive:
In the European Union, this product will be considered Waste Electric and Electronic Equipment (WEEE) at the end of its life.

recommended

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.

Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.

Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.

Discover the benefits of MILLIPLEX® multiplex assays, based on Luminex® xMAP® multiplex assay technology, that provide consistent, high-quality results and see how these multiplex biomarker Luminex® assays are being used to advance research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service