Skip to Content
MilliporeSigma
All Photos(1)

Documents

HX0299

Supelco

Hexane, mixture of isomers

≥98.5% (total hexane isomers + methylcyclopentane)

Synonym(s):

Hexanes, FM approval

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6

product name

Hexanes, 98.5% ACS, ~64% n-Hexane, Meets ACS Specifications

vapor pressure

160 hPa ( 20 °C)

Quality Level

Assay

≥98.5% (total hexane isomers + methylcyclopentane)

form

liquid

impurities

≤0.0003 meq/g Water-soluble titrable acid
≤0.005% Sulfur compounds (as S)

evapn. residue

≤0.001%

color

APHA: ≤10

bp

69 °C/1013 hPa

mp

-94.3 °C

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.66 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The mixture of hexane isomers and methylcyclopentane can be used as a solvent in the silica gel adsorption chromatographic method for the purification of organic compounds.

Analysis Note

Assay (n-hexane): Report%
Assay (total hexane isomers + methylcyclopentane): 98.5% min
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Sulfur compounds (as S): 0.005% max
Thiophene: To pass test
Water-soluble titrable acid: 0.0003meq/g max

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles Rubert Pérez et al.
Chemical biology & drug design, 79(3), 260-269 (2011-12-08)
Two molecular scaffolds were designed using the CAVEAT molecular design package to inhibit the oligomerization of protective antigen (PA(63) ), a key protein component of anthrax toxin. The inhibitors were designed to prevent heptamerization of PA(63) by mimicking key
Ring and side chain reactivities of 1-([1, 3, 4] oxadiazol-2-ylmethyl)-1H-benzotriazoles
Katritzky AR, et al.
ARKIVOC (Gainesville, FL, United States), 2 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service