Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HX0295

Supelco

Hexane

≥95.0% (n-hexane basis), suitable for LC/MS, gas chromatography (GC) and HPLC, OmniSolv®

Synonym(s):

Hexane, n-Hexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH3
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-777-6
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
application(s):
PFAS testing
bp:
69 °C (lit.)
vapor pressure:
256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)
Pricing and availability is not currently available.

Product Name

n-Hexane 95%, OmniSolv®

vapor density

~3 (vs air)

Quality Level

vapor pressure

256 mmHg ( 37.7 °C)
5.2 psi ( 37.7 °C)
~132 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥95.0% (n-hexane basis)

form

liquid

autoignition temp.

453 °F

expl. lim.

7.7 %

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)
≤3 μg/L Capillary FID responsive substances (as decane)
≤5 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.375 (lit.)

bp

69 °C (lit.)

mp

−95 °C (lit.)

transition temp

flash point -22 °C

solubility

0.0095 g/L

density

0.659 g/mL at 25 °C (lit.)

UV cutoff

≤195 nm

UV absorption

λ: 195 nm Amax: ≤1.00
λ: 200 nm Amax: ≤0.50
λ: 210 nm Amax: ≤0.20
λ: 220 nm Amax: ≤0.05
λ: 230 nm Amax: ≤0.05
λ: 250 nm Amax: ≤0.01
λ: 280 nm Amax: ≤0.005

application(s)

PFAS testing

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCC

InChI

1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3

InChI key

VLKZOEOYAKHREP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

HX0295P-1 is packaged in poly coated glass bottles
OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (n-hexane)95% minBoiling range66.1-69.4°CCapillary ECD responsive substances (as PCNB)5ng/L maxCapillary FID responsive substances (as decane)3µg/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 195 nm1.00AU maxUV Abs. at 200 nm0.50AU maxUV Abs. at 210 nm0.20AU maxUV Abs. at 220 nm0.05AU maxUV Abs. at 230 nm0.05AU maxUV Abs. at 250 nm0.01AU maxUV Abs. at 280 nm0.005AU maxUV Cut-off195nm maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service