Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HNDG4MAG-36K

Millipore

MILLIPLEX® Human Neurodegenerative Disease Magnetic Bead Panel 4 - Neuroscience Multiplex Assay

The analytes available for this multiplex kit are: Amyloid β40, Amyloid β42, GDNF, sRAGE, S100B.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 84-108%
standard curve range: 14-10,000 pg/mL
(S100B)

standard curve range: 21-15,000 pg/mL
(sRAGE)

standard curve range: 27-20,000 pg/mL
(Amyloid β42)

standard curve range: 3-2,500 pg/mL
(Amyloid β40)

standard curve range: 7-5,000 pg/mL
(GDNF)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

The MILLIPLEX® Human Neurodegenerative Bead Panel 4 is to be used for the simultaneous quantification of the following 5 analytes in any combination in human cerebrospinal fluid (CSF): S100B, Amyloid β40 (Aβ1-40), Amyloid β42 (Aβ1-42), sRAGE, and GDNF. This kit is not validated for testing serum or cell culture supernatants.

Panel Type: Neuroscience

Specificity

Cross Reactivty
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: Amyloid β40, Amyloid β42, GDNF, sRAGE, S100B
  • Recommended Sample type: CSF
  • Recommended Sample dilution: 1:3
  • Assay Run Time: Overnight
  • Research Category: Neuroscience

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Packaging

Everything you need in a single kit.

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual analytes.

Legal Information

MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Justice et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(6), 2612-2623 (2015-02-13)
Recent studies have found that those who suffer from posttraumatic stress disorder (PTSD) are more likely to experience dementia as they age, most often Alzheimer's disease (AD). These findings suggest that the symptoms of PTSD might have an exacerbating effect

Related Content

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

Neuroscience multiplex assays measure biomarkers to understand neurodegenerative diseases, disorders, and neuropeptide signaling.

Multiplex detection of Alzheimer's biomarkers with MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel aids research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service