Skip to Content
MilliporeSigma
All Photos(1)

Documents

HIFN-130K

Millipore

MILLIPLEX® Human Interferon Panel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.77

usage

 kit sufficient for 96 wells

Quality Level

species reactivity

human

parameter

25 μL sample volume (per well)

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

Interferons (IFNs) are widely expressed cytokines that have potent antiviral and antiproliferative effects. These cytokines are the first line of defense against viral infections and have important roles in immunosurveillance for malignant cells. IFNs are classified as type I, II and III IFNs, based upon the target receptors to which they bind. In humans, type I IFNs belong to a multigene family consisting of multiple IFNα subtypes but only one IFNβ, IFNε, IFNκ, and IFNω. There is only one type II IFN: IFNγ. The type III IFNs include IFNλ1, λ2 and λ3, which are also known as interleukin-29 (IL-29), IL 28A and IL-28B, respectively. Functionally, the type I and type III IFNs are induced during a viral infection and involved in host defense against viruses. IFNγ (type II) is primarily involved in the allergic response, in host defense against intracellular pathogens, and in control of tumor growth. Studies of IFNs have broadened the mechanistic understanding of cell signaling, gene transcription, and innate and acquired immunity.

Research category: Inflammation & ImmunologyResearch sub-category:

  • Inflammation & Autoimmune Mechanisms
  • Inflammation

Specificity

There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

The MILLIPLEX® Human Interferon Magnetic Bead Panel is a 9-plex kit to be used for the simultaneous quantification of any or all of the following analytes in serum or plasma samples and tissue/cell lysate and culture supernatant samples: IFNα-2, IFNβ, IFNε, IFNɣ, IFNɣ Receptor 1, IFNλ1, IFNλ2, IFNλ3, IFNω.

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

Guide on selecting immunoassay platforms including multiplex vs. singleplex assays, instruments, software, and services.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service