Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HC5B9-140K

Millipore

MILLIPLEX® Human Complement C5b-9 - Singleplex

Synonym(s):

Human complement multiplex kit, Luminex human complement immunoassay, Millipore human complement panel, Human membrane attack complex,, human MAC panel, Luminex human complement panel, terminal complement complex, MAC, membrane attack complex

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

species reactivity

human

Quality Level

parameter

25 μL sample volume (per well diluted sample)

assay range

recovery range: 100%
(same day assay)

recovery range: 120%
(overnight assay)

inter-assay cv: ≤15%
intra-assay cv: ≤15%
standard curve range: 0.6-450 ng/mL

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

shipped in

wet ice

storage temp.

2-8°C

General description

The complement system consists of many plasma proteins that assist, or complement, the ability of phagocytic cells and antibodies to clear pathogens. Although it is part of the innate immune system, it can be recruited and activated by the adaptive immune system. The complement proteins and glycoproteins that are synthesized primarily by hepatocytes normally circulate as inactive pro-proteins. Once stimulated the result of the cascade is a massive amplification of response and activation of the membrane attack complex (C5b-9), which forms a transmembrane pore causing osmotic lysis of target cells. The complement pathway may also form soluble C5b-9, as well as intermediate isoforms, stabilized by regulatory proteins. These soluble complexes are unable to insert into the lipid membrane, and their role in complement pathways is an area of ongoing research.

Research category : Inflammation & Immunology
Research sub-category : Inflammation & Autoimmune Mechanisms; Inflammation

Specificity

Cross-Reactivity between C5b-9 and other complement proteins found in MILLIPLEX® Human Complement Expanded panel 1 (Cat. No. HCMPEX1-19K) and MILLIPLEX® Human Complement Panel 2 (Cat. No. HCMP2MAG-19K) was tested. There was no or negligible cross-reactivity between the antibody pair in this panel for any of the analytes tested.

Application

MILLIPLEX® Human Complement C5b-9 – Singleplex Magnetic Bead Panel is a Singleplex kit to be used for the quantification of the following analyte in serum, plasma, or cell culture supernatant samples: C5b-9 (Membrane Attack Complex, Terminal Complement Complex).

Features and Benefits

Everything you need in a single kit

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore how to select the best immunoassay platform for your research by choosing between multiplex and singleplex biomarker detection assays, immunoassay instrument platforms, immunoassay software, and services.

Explore how to select the best immunoassay platform for your research by choosing between multiplex and singleplex biomarker detection assays, immunoassay instrument platforms, immunoassay software, and services.

Explore how to select the best immunoassay platform for your research by choosing between multiplex and singleplex biomarker detection assays, immunoassay instrument platforms, immunoassay software, and services.

Explore how to select the best immunoassay platform for your research by choosing between multiplex and singleplex biomarker detection assays, immunoassay instrument platforms, immunoassay software, and services.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service