Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FR139430087

Sigma-Aldrich

Estapor®White Microspheres

K-020 White Std PS 10%, White microspheres 200nm, Latex Turbidimetric Immunoassay - For ordering, click "Request more information"

Synonym(s):

plain beads, white beads, white particles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13102026
NACRES:
NA.24

material

high-density polyethylene packaging (bottle)
polystyrene beads

Quality Level

manufacturer/tradename

Estapor®

packaging

bottle of 10ml-10L

concentration

10% solids

technique(s)

latex turbidmetry immunoassay (LTIA): suitable

color

white

matrix

polymeric polystyrene

diameter

0.176 - 0.225 μm±<5%

functional group

(plain)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Estapor® standard polystyrene microspheres offer a simple and convenient option for the development of a wide range Latex Turbidimetry Immunoassay (LTIA) diagnostic tests. The plain surface of the microsphere is designed to be easily and quickly coated with the antibody or antigen of choice via protein adsorption through hydrophobic and ionic interactions. The simplicity of this conjugation process is a key feature of these microspheres and has attracted many diagnostic test developers whom have successfully utilized these microspheres for several years in a wide variety of LTIAs.

Application

Main applications of the standard microspheres include nephelemetric assays, immune-turbidimetric assays, latex agglutination tests, solid phase immunoassays, microspheres capture ELISA, calibration

Legal Information

Estapor is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Estapor® Microspheres, for polymeric supports in diagnostics, life sciences, biotechnology, & environmental applications, offers 100+ microsphere types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service