Skip to Content
MilliporeSigma

I9016

Anti-Interferon-γ antibody produced in goat

IgG fraction of antiserum

Synonym(s):

Anti-IFN-γ

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
EC Number:
202-739-6
NACRES:
NA.41
MDL number:
Conjugate:
unconjugated
Clone:
polyclonal
Application:
Neutral, WB
Citations:
5
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

neutralization: suitable, western blot: suitable

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFNG(3458)

General description

Interferons (IFNs) are cytokines that are secreted in response to viral or bacterial infections and tumorigenesis. There are two types of IFNs, type I (IFN-α and IFN-β) and type II (IFN-γ). IFN-γ binds to specific receptor complex consisting of IFNγR1 and IFNγR2. There are many pathways that are mediated by IFN-γ binding such as JAK/STAT1, AP-1, NF-κB, STAT3 and STAT5. IFN-γ is pleotropic and performs various functions related to immune response, inflammation, differentiation and activation of T cells, cell cycle and apoptosis. The most studied function of IFN-γ is priming the antigen presenting cells by upregulating major histocompatibility (MHC) Class I molecules. IFN-γ has clinical applications in autoimmune diseases (rheumatoid arthritis), multiple sclerosis, cancer, HIV and fungal infections
Anti-Interferon-γ recognizes human Interferon-γ. It does not bind specifically to mouse, hamster or bovine IFN-γ.

Immunogen

recombinant human IFN-γ.

Application

Anti-interferon-γ antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. The antibody is suitable for neutralization reactions.

Biochem/physiol Actions

The antibody shows no cross-reactivity with recombinant mouse IFN-γ.

Physical form

Lyophilized from 0.2 um-filtered solution in PBS, pH 7.4 with 5% Trehalose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library