Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FE-125ML-BK

Millipore

Intermediates For Further Manufacturing Use (FFMU) - Rare Specificities

Anti-s from the clone P3BER, Human IgM, Intermediate for further manufacturing use

Synonym(s):

Anti-s (clones MNS4/P3BER), FFMU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

biological source

human

Quality Level

antibody form

unpurified

clone

P3BER, monoclonal

form

liquid

packaging

pkg of 125 mL

concentration

10-25 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6.7-7.7

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

General description

Standardized, concentrated cell culture supernatant which has been diafiltered with phosphate buffered saline.; For the formulation of Anti-s blood typing reagents or devices.; Contains 0.1% (w/v) Sodium Azide; Shelf Life: 18 months from date of filtation.

Application

Antibody intermediate for further manufacturing use
Production of an in vitro blood typing product or device

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Partially processed reagent for further manufacture of non-injectable products only. The absence of all viral antigens has not been determined. No US Standard of Potency. Do not use if turbid. Contains 0.1% (w/v) sodium azide.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service