Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GSWP01300

Millipore

MCE Membrane Filter, 0.22 μm Pore Size

MF-Millipore, filter diam. 13 mm, hydrophilic, white

Synonym(s):

MF-Millipore Membrane Filter, 0.22 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Pricing and availability is not currently available.

material

mixed cellulose esters (MCE) membrane
plain filter
white filter

Quality Level

description

13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic, white, 100 discs

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

MF-Millipore
Millipore

parameter

18 mL/min-cm2 water flow rate
2 L/min-cm2 air flow rate
75 °C max. temp.

diam.

13 mm

filter diam.

13 mm

thickness

150 μm

gravimetric extractables

2%

color

white

refractive index

n/D 1.51

matrix

MF-Millipore

pore size

0.22 μm pore size
75 % porosity

capacity

150 μg/cm2 binding capacity (protein)

bubble point

≥3.52 bar, air with water at 23 °C

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Application

  • Sterilizing filtration
  • Bioassays
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Other Notes

Directions for Use

  • Organism Retention: Microorganism
  • Mode of Action: Filtration (size exclusion)
  • Application: General laboratory filtration
  • Intended Use: Retention or removal of biological contaminants
  • Instructions for Use: Sterilizing filtration of a liquid through a membrane with a 0.2 μm (or smaller) pore size effectively removes biological contaminants, including bacteria, mold and yeast. For the selective retention of larger biological contaminents, liquid filtration through membranes with 0.45 μm (or larger) pore sizes may be used to trap and support microorganism growth for subsequent culture and analysis
  • Storage Statement: Store in dry location away from heat source
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroyuki Imachi et al.
Nature, 577(7791), 519-525 (2020-01-17)
The origin of eukaryotes remains unclear1-4. Current data suggest that eukaryotes may have emerged from an archaeal lineage known as 'Asgard' archaea5,6. Despite the eukaryote-like genomic features that are found in these archaea, the evolutionary transition from archaea to eukaryotes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service