Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB203P

Sigma-Aldrich

Milli-Mark® Anti-CD64-PE Antibody, clone 10.1

clone 10.1, Milli-Mark®, from mouse

Synonym(s):

High affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I, Fc-gamma RIA, FcgammaRIa, Fc-gamma RI, FcRI

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

10.1, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FCGR1A(2209)

Related Categories

General description

CD64 is the high-affinity IgG Fc receptor belonging to the immunoglobulin supergene family (Guyre, 2002). CD64 is a 72 kDa glycoprotein which binds monomeric IgG (human IgG>>IgG>>>IgG4>>>IgG2 and mouse IgG3>IgG2a>>>IgG1, IgG2b). Three genes: FcγR1A, FcγR1B and FcγR1C, which have >98 percent identity at the nucleotide level, have been identified and mapped to chromosome 1, band q21.1 (Sun,1997)). The FcγR1A gene, which specified the high-affinity IgG receptor, encodes three C2-type Ig-like extracellular domains, a 21 amino acid transmembrane region, and a charge cytoplasmic tail of 61 amino acids (Sun, 1997)(van de Winkel,1993).

Specificity

Antibody recognizes Human CD64 antigen.

Immunogen

Human rheumatoid synovial fluid cells and monocytes.

Application

Milli-Mark Anti-CD64-PE Antibody, clone 10.1 detects level of CD64 & has been published & validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human PBMCs

Target description

72 kDa (Sun W, 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMC′s

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service