Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DX1732

Supelco

N,N-Dimethylformamide

Biosynthesis OmniSolv®

Synonym(s):

Formic Acid Dimethylamide, DMF, DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-679-5
application(s):
peptide synthesis
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)

vapor density

2.5 (vs air)

Quality Level

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

833 °F

shelf life

24 mo. (from manufacture date)

expl. lim.

15.2 %

dilution

(for synthesis)

impurities

≤0.4 μeq/g Titrable acid
≤0.4 μeq/g Titrable base
≤200 ppm Water (H2O)
≤5 ppm Amines (as dimethylamine)

evapn. residue

≤5 ppm

color

APHA: ≤10

refractive index

n20/D 1.430 (lit.)

pH

7 (20 °C, 200 g/L in H2O)

bp

153 °C (lit.)

mp

−61 °C (lit.)

transition temp

flash point 57.5 °C

density

0.944 g/mL (lit.)

UV cutoff

≤270 nm

UV absorption

λ: 270 nm Amax: ≤1.00
λ: 275 nm Amax: ≤0.30
λ: 280 nm Amax: ≤0.20
λ: 300 nm Amax: ≤0.05
λ: 350 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.005

application(s)

peptide synthesis

shipped in

ambient

storage temp.

room temp

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biosynthesis OmniSolv®
Solvent for many hydrophobic organic compounds.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Expiration dateExp 24 months from mfg dateAmines (as dimethylamine)5ppm maxAssay (GC)99.9% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.4µeq/g maxTitrable base0.4µeq/g maxUV Abs. at 270 nm1.00AU maxUV Abs. at 275 nm0.30AU maxUV Abs. at 280 nm0.20AU maxUV Abs. at 300 nm0.05AU maxUV Abs. at 350 nm0.01AU maxUV Abs. at 400 nm0.005AU maxUV Cut-off270nm maxWater (H2O)200ppm max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service