Skip to Content
MilliporeSigma
All Photos(4)

Documents

918660

Sigma-Aldrich

γ-Valerolactone

greener alternative

BioRenewable, ≥99%, ReagentPlus®

Synonym(s):

GVL, γ-Methyl-γ-butyrolactone, 4,5-Dihydro-5-methyl-2(3H)-furanone, 4-Hydroxypentanoic acid lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O2
CAS Number:
Molecular Weight:
100.12
Beilstein:
80420
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

vapor density

3.45 (vs air)

Quality Level

product line

ReagentPlus®

Assay

≥99%

form

liquid

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Design for Degradation
Learn more about the Principles of Green Chemistry.

refractive index

n20/D 1.432 (lit.)

bp

207-208 °C (lit.)
82-85 °C/10 mmHg (lit.)

mp

−31 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CC1CCC(=O)O1

InChI

1S/C5H8O2/c1-4-2-3-5(6)7-4/h4H,2-3H2,1H3

InChI key

GAEKPEKOJKCEMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” ,“Use of Renewable Feedstock” and "Design for Degradation".

Application

Cross-coupling reactions
  • HATU Amide Coupling
  • Suzuki-Miyaura coupling reaction
  • Sonogashira coupling reaction





ATRP polymerization
  • Replacement of DCM in peptide synthesis




Features and Benefits

  • Made from Renewable Resource - rice
  • Biodegradable
  • Non-toxic
  • Low vapor pressure

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service