Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DX1574

Sigma-Aldrich

Dimethylamine

40% in Water

Synonym(s):

Dimethylamine solution, DMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
MDL number:
UNSPSC Code:
12352116

vapor pressure

263 hPa ( 20 °C)

form

liquid

pH

14 ( in H2O)

mp

-38 °C

transition temp

flash point -18.5 °C

shipped in

ambient

storage temp.

room temp

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Appearance: Colorless to pale straw colored
Assay: 40% min
Form: Clear liquid

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-1.3 °F

Flash Point(C)

-18.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service