Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CBL194F

Sigma-Aldrich

Anti-Cytokeratin 7 Antibody, clone LP5K, FITC conjugated

clone LP5K, Chemicon®, from mouse

Synonym(s):

Cytokeratin 7 Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

LP5K, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... KRT7(3855)

Specificity

The antibody reacts with cytokeratin 7 which has been found in columnar and glandular epithelium of the lung, cervix, breast, bile ducts and larger collecting ducts of the kidney. Occasionally staining of blood vessel cell walls, particularly endothelial cells, may be observed.

FUSION PARTNER: Sp2/0

Immunogen

Cytoskeletal proteins from cultured HeLa cells

Application

Detect Cytokeratin 7 using this Anti-Cytokeratin 7 Antibody, clone LP5K, FITC conjugated validated for use in IC, IH, IH(P), FC.
Immunohistochemistry: Suitable for staining frozen sections and paraffin wax embedded tissue. Protease treatment of paraffin embedded sections is required prior to antibody application. Frozen sections (7 µm) of placental tissue fixed with acetone for 1 hr. Antibody used at 1:10 dilution (Sato et al., 2003).

Immunocytochemistry: isolated EVT cells fixed with 0.5% PFA at 4°C for 15 minutes followed by acetone at -20°C for 2 minutes. Antibody was used at 1:10 dilution (Sato et al., 2003).

Flow cytometry: 10μL of conjugate per test.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
Cytokeratins

Quality

Tested

Physical form

The antibody is ammonium sulphate precipitation followed by ion exchange chromatography. The conjugate is supplied in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin.

Storage and Stability

Store at +4°C protected from light. DO NOT FREEZE. For long term use and storage aliquot conjugate into small volumes and store at +4°C for up to one year.

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silvia Giugliano et al.
Journal of immunology (Baltimore, Md. : 1950), 195(8), 3737-3747 (2015-09-06)
Hepatitis C virus (HCV) is the world's most common blood-borne viral infection for which there is no vaccine. The rates of vertical transmission range between 3 and 6% with odds 90% higher in the presence of HIV coinfection. Prevention of
Zhiyuan Hu et al.
Cancer cell, 37(2), 226-242 (2020-02-13)
The inter-differentiation between cell states promotes cancer cell survival under stress and fosters non-genetic heterogeneity (NGH). NGH is, therefore, a surrogate of tumor resilience but its quantification is confounded by genetic heterogeneity. Here we show that NGH in serous ovarian cancer
Amandine Vargas et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 28(8), 3703-3719 (2014-05-09)
Exosomes are extracellular vesicles that mediate intercellular communication and are involved in several biological processes. The objective of our study was to determine whether endogenous retrovirus group WE, member l (ERVWE1)/syncytin-1 and endogenous retrovirus group FRD, member 1 (ERVFRDE1)/syncytin-2, encoded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service