Skip to Content
MilliporeSigma
All Photos(1)

Documents

BX0220

Supelco

Benzene

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Benzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
MDL number:
UNSPSC Code:
12352005
EC Index Number:
200-753-7

Agency

meets USP/NF

Quality Level

vapor pressure

101 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

impurities

≤0.005% Sulfur compounds (as S)
≤0.05% Water (H2O)

evapn. residue

≤0.001%

color

APHA: ≤10

bp

80.1 °C/1013 hPa

mp

5.5 °C

transition temp

flash point -11 °C

solubility

1.88 g/L

density

0.88 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

General description

Benzene (C6H6) is an organic compound. It is widely used as a solvent in the production of industrial chemicals (such as cyclohexane, cumene, and styrene), and adhesives. Additionally, several types of rubber, lubricants, dyes, detergents, drugs, and pesticides are made using benzene.

Application

Benzene can be used:
  • As a starting material to produce ethylbenzene with the gaseous ethylene in presence of Friedel-Crafts catalysts (aluminum chloride).
  • As a starting material to produce cumene by catalytic alkylation with propylene.
  • As a reactant in the production of cyclohexane under mild condition using Ru/ carbon nanotubes (CNTs) catalysts by hydrogenation method.
  • To synthesis phenol using H2O2 as an oxygen source and nickel(II)complex as a catalyst by hydroxylation method.
  • As a solvent along with hexafluoro benzene to obtain stabilized graphene sheet.

Analysis Note

Assay (GC): 99.0% min
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Substances darkened by sulfuric acid: To pass test
Sulfur compounds (as S): 0.005% max
Thiophene: To pass test (limit 1 ppm)
Water (H2O): 0.05% max

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service