Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX0212

Supelco

Benzene

OmniSolv®

Synonym(s):

Benzene, Benzine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
MDL number:
UNSPSC Code:
12352005
EC Index Number:
200-753-7
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
80.1 °C/1013 hPa
vapor pressure:
101 hPa ( 20 °C)

vapor pressure

101 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99.7% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.03% Water (H2O)

evapn. residue

≤2 ppm

color

APHA: ≤10

bp

80.1 °C/1013 hPa

mp

5.5 °C

transition temp

flash point -11 °C

solubility

1.88 g/L

density

0.88 g/cm3 at 20 °C

UV cutoff

≤280 nm

UV absorption

λ: 280 nm Amax: ≤1.00
λ: 290 nm Amax: ≤0.15
λ: 300 nm Amax: ≤0.05
λ: 330 nm Amax: ≤0.01
λ: 350 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Application

OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.7% minColor (APHA)10 maxEvaporation residue2ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 280 nm1.00AU maxUV Abs. at 290 nm0.15AU maxUV Abs. at 300 nm0.05AU maxUV Abs. at 330 nm0.01AU maxUV Abs. at 350 nm0.005AU maxUV Cut-off280nm maxWater (H2O)0.03% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service