Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BI0424

Sigma-Aldrich

Oxidizing Reagent, 0.05M, I₂ in Pyridine

Oxidizing Reagent, 0.05M, I₂ in Pyridine

Biosynthesis Novabiochem®

Synonym(s):

dna

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116006
Pricing and availability is not currently available.

Quality Level

product line

Novabiochem®

form

liquid

reaction suitability

reaction type: Oxidations
reagent type: oxidant

transition temp

flash point 20 °C

shipped in

ambient

storage temp.

2-30°C

General description

I₂ in Pyridine is an oxidizing agent that are appropriate for all types of biosynthetic processes, including the synthesis of DNA, RNA, and peptides.

Application

BI0424-1005 is packaged in GL45 glass bottles
BI0424-2505 is packaged in GL45 glass bottles
BI0424-P4000 is packaged in poly coated glass bottles
BI0424-4000 is packaged in 38/430 glass bottles
DNA Reagents Label Change Explanation
Biosynthesis Novabiochem®

Analysis Note

% Pyridine concentration: 89.0-91.0%
Color: Dark brown
Component: Pyridine Purity: 99.0% min
Form: Liquid
Iodine Molarity: 0.04-0.06M
Water (H2O): 9.0-11.0%

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service