Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP500

Sigma-Aldrich

Goat Anti-Mouse IgM µ chain Antibody, Species Adsorbed

1 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

General description

IgM constitutes about 10% of serum immunoglobulins. IgM antibody is prominent in early immune responses to most antigens and predominates in certain antibody responses such as ′natural′ blood group antibodies. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. The gene for the mu constant region contains four domains separated by short intervening sequences.

Specificity

Reacts with heavy chain of mouse IgM. Absorbed for mouse IgG1, IgG2a, IgG2b, IgG3 and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins. Reactivity with other species has not been determined.

Application

This Goat anti-Mouse IgM µ chain Antibody, Species Adsorbed is validated for use in ELISA for the detection of Mouse IgM µ chain.

Physical form

Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 1.0 mL of 100 mM borate buffered saline, pH 8.2.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Schultheiß et al.
Science advances, 10(34), eadl3975-eadl3975 (2024-08-21)
Genetic TNFAIP3 (A20) inactivation is a classical somatic lymphoma lesion and the genomic trait in haploinsufficiency of A20 (HA20). In a cohort of 34 patients with HA20, we show that heterozygous TNFAIP3 loss skews immune repertoires toward lymphocytes with classical
Seung-Chul Choi et al.
Journal of immunology (Baltimore, Md. : 1950), 208(9), 2098-2108 (2022-04-08)
Several studies have shown an enhanced metabolism in the CD4+ T cells of lupus patients and lupus-prone mice. Little is known about the metabolism of B cells in lupus. In this study, we compared the metabolism of B cells between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service