Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP128F

Sigma-Aldrich

Goat Anti-Mouse IgM Antibody, µ chain, FITC conjugate

2 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Based on immunoelectrophoresis, the antibody reacts with the heavy chain portion of IgM. No reactivity with IgG or non-immunoglobulin serum proteins was detected, but cross reactivity with IgM from other species may be detected.

F/P RATIO:

9.0 mg/mg; 3.4 moles FITC per mole IgG

Application

Goat anti-Mouse IgM Antibody, μ chain, FITC conjugate detects level of Mouse IgM & has been published & validated for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Physical form

Freeze dried powder. Leyofilized from liquid containing 0.01M sodium phosphate, pH7.6, 0.25M sodium chloride, 15 mg/mL bovine serum albumin, and 0.05% sodium azide.

RECONSTITUTION:

Reconstitute in 1mL deionized water.
ImmunoAffinity Purified

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Peng et al.
OncoTargets and therapy, 13, 8677-8689 (2020-09-29)
Thomsen-Friedenreich antibody (TF-Ab) is a specific antibody against the Thomsen-Friedenreich antigen (TF-Ag). At present, studies on a number of other tumors have shown that TF-Ab can effectively inhibit metastasis and induce apoptosis in tumor cells. However, the role of TF-Ab
Francesca Cialdai et al.
Cellular and molecular life sciences : CMLS, 78(23), 7795-7812 (2021-10-30)
Astronauts on board the International Space Station (ISS) are exposed to the damaging effects of microgravity and cosmic radiation. One of the most critical and sensitive districts of an organism is the eye, particularly the retina, and > 50% of astronauts develop
Katsuyoshi Kumagai et al.
Experimental animals, 66(2), 125-136 (2016-11-29)
Targeted mutant mice generated on a C57BL/6 background are powerful tools for analysis of the biological functions of genes, and gene targeting technologies using mouse embryonic stem (ES) cells have been used to generate such mice. Recently, a bacterial artificial
Jamie L Marshall et al.
Human molecular genetics, 21(20), 4378-4393 (2012-07-17)
Sarcospan (SSPN) is a core component of the major adhesion complexes in skeletal muscle, the dystrophin- and utrophin (Utr)-glycoprotein complexes (DGC and UGC). We performed a rigorous analysis of SSPN-null mice and discovered that loss of SSPN decreased DGC and
M S Yoon et al.
European journal of pain (London, England), 22(9), 1691-1700 (2018-06-05)
Neuropathic pain is associated with abnormal sensitivity of the central nervous system. Although the mechanism underlying the development of sensitization remains to be fully elucidated, recent studies have reported that neuroplastic changes in the pain circuitry may be involved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service