Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

ABS554

Sigma-Aldrich

Anti-phospho-FKHRL1/FOXO3A (Thr32)

from rabbit, purified by affinity chromatography

Synonym(s):

Forkhead box protein O3, AF6q21 protein, Forkhead in rhabdomyosarcoma-like 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (pThr32)

Gene Information

human ... FOXO3(2309)

General description

Forkhead box protein O3 (UniProt: O43524; also known as AF6q21 protein, Forkhead in rhabdomyosarcoma-like 1) is encoded by the FOXO3 (also known as FKHRL1, FOXO3A) gene (Gene ID: 2309) in human. Forkhead box protein O3 is a transcriptional activator that riggers apoptosis in the absence of survival factors, including neuronal cell death upon oxidative stress. It recognizes and binds to the DNA sequence 5′-[AG]TAAA[TC]A-3′. Its DNA binding domain located in amino acids 157-251. The nuclear localization signal is located in amino acids 242-259. In the cytoplasm it is present in an inactive state and it translocates to the nucleus upon oxidative stress and in the absence of survival factors. In the presence of survival factors such as IGF-1, it is phosphorylated on Thr32 and Ser253 by AKT1/PKB. The phosphorylated form then interacts with 14-3-3 proteins and is retained in the cytoplasm. Survival factor withdrawal induces dephosphorylation and promotes translocation to the nucleus where the dephosphorylated protein induces transcription of target genes and triggers apoptosis. Forkhead box protein O3 deacetylation by SIRT1 or SIRT2 stimulates its interaction SKP2 and facilitates SCF(SKP2)-mediated FOXO3 ubiquitination and proteasomal degradation. A chromosomal aberration involving FOXO3 is found in secondary acute leukemias.

Specificity

This rabbit polyclonal antibody detects Forkhead box protein O3 in human cells. It targets an epitope within 11 amino acids surrounding phosphorylated Thr32 from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphorylated Thr32 from the N-terminal region of human Forkhead box protein O3.

Application

Anti-phospho-FKHRL1/FOXO3A (Thr32), Cat. No. ABS554, is a rabbit polyclonal antibody that detects Forkhead box protein O3 and has been tested for use in Immunohistochemistry (Paraffin), Peptide Inhibition Assay, and Western Blotting.
Peptide Inhibition Assay: A 1:500 dilution from a representative lot blocked signal in HEK293 cells treated with IGF.

Immunohistochemistry Analysis: A 1:150 dilution from a representative lot detected phospho-FKHRL1/FOXO3A (Thr32) in human tonsil, human small intestine, and human liver cancer tissues.
Research Category
Signaling

Quality

Evaluated by Western Blotting in HEK293 treated with IGF stimulation cell lysate.

Western Blotting Analysis: A 1:100 dilution of this antibody detected phospho-FKHRL1/FOXO3A (Thr32) in lysate from HEK293 cells treated with IGF (25 ng/mL for 10 min.).

Target description

~75 kDa observed; 71.28 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linxi Lv et al.
Translational oncology, 46, 101989-101989 (2024-05-24)
Lung cancer has one of the highest mortality rates worldwide, with non-small-cell lung cancer (NSCLC) constituting approximately 85% of all cases. Demethylzeylasteral (DEM), extracted from Tripterygium wilfordii Hook F, exhibits notable anti-tumor properties. In this study, we revealed that DEM
Hao Liu et al.
Oncogenesis, 11(1), 19-19 (2022-04-24)
Previously, our lab explored that tongue cancer resistance-associated protein (TCRP1) plays a central role in cancer chemo-resistance and progression. Absolutely, TCRP1 was significantly increased in lung cancer. But the mechanism is far from elucidated. Here, we found that TCRP1 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service