Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABS47

Sigma-Aldrich

Anti-phospho-SMAD3 (Tyr179) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Mothers against DPP homolog 3, MAD homolog 3, Mad3, Mothers against decapentaplegic homolog 3, JV15-2, AMAD3, SMAD family member 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine, bovine, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr179)

Gene Information

human ... SMAD3(4088)

General description

Members of the Smad family of signal transduction molecules are components of a critical intracellular pathway that transmits TGF-β signals from the cell surface into the nucleus. Three distinct classes of Smads have been defined: the receptor-regulated Smads (R-Smads), which include Smad1, 2, 3, 5 and 8, the common-mediator Smad (co-Smad), Smad4, and the antagonistic or inhibitory Smads (I-Smads), Smad6 and 7. Once in the nucleus, Smads can target a variety of DNA binding proteins to regulate transcriptional responses. Following stimulation by TGF-β, Smad2 and Smad3 become phosphorylated at their carboxyl termini (Ser465 and 467 on Smad2; Ser423 and 425 on Smad3) by TGF-β Receptor I. Phosphorylated Smad 2/3 can complex with Smad4, translocate to the nucleus and regulate gene expression.

Specificity

Other homologies: Zebrafish (91% sequence homology), Chicken (91% sequence homology) and Xenopus (91% sequence homology).
This antibody recognizes the Linker region of SMAD3 phosphorylated at Thr179.

Immunogen

Epitope: Linker region
KLH-conjugated linear peptide corresponding to the Linker Region of human SMAD3 phosphorylated at Tyr179..

Application

Detect phospho-SMAD3 (Tyr179) using this Anti-phospho-SMAD3 (Tyr179) Antibody validated for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in untreated and TGF Beta-treated HaCat cell lysates.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SMAD3 in 10 mg of untreated and TGF Beta-treated HaCat cell lysates.

Target description

~52 kDa observed. Uncharacterized bands at ~170 kDa, ~165 kDa, ~90 kDa and ~52 kDa may be observed in untreated HaCat cell lysate and in some other cell lysates. Uncharacterized bands at ~105 kDa and ~55 kDa may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.5% sodium azide.

Storage and Stability

Stable for 1 year at 2-8º from date of receipt.

Analysis Note

Control
Untreated and TGF Beta-treated HaCat cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service