Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3849-I

Sigma-Aldrich

Anti-phospho SMAD2 Antibody (Ser465/467)

from rabbit, purified by affinity chromatography

Synonym(s):

Mothers against decapentaplegic homolog 2, Ser465/467 phosphorylated, hMAD-2, Ser465/467 phosphorylated, hSMAD2, Ser465/467 phosphorylated, JV18-1, Ser465/467 phosphorylated, MAD homolog 2, Ser465/467 phosphorylated, Mad-related protein 2, Ser465/467 pho

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

zebrafish (based on 100% sequence homology), Xenopus (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology), chicken (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer465/pSer467 )

Gene Information

human ... SMAD2(4087)

General description

Mothers against decapentaplegic homolog 2 (UniProt Q15796; also known as hMAD-2, hSMAD2, JV18-1, MAD homolog 2, Mad-related protein 2, Mothers against DPP homolog 2, SMAD 2, SMAD family member 2, Smad2) is encoded by the SMAD2 (also known as MADH2, MADR2) gene (Gene ID 4087) in human. Smad2 is a receptor-regulated intracellular signal transducer and transcriptional modulator activated by transforming growth factor TGF-beta and activin type 1 receptor kinases. In response to TGF-beta signal, smad2 is recruited to the activated TGF-beta receptor through its interaction with the SMAD anchor for receptor activation (SARA) protein, where it is phosphorylated by the TGF-beta receptor. The phosphorylation results in its dissociation from SARA and association with another family member smad4. The smad2-smad4 complex then translocates into the nucleus, where it binds to target promoters and forms a transcription repressor complex with other cofactors. Smad2 can also be phosphorylated by activin type 1 receptor kinase, and thereby transduce activin-dependent signaling events.

Specificity

This antibody detects Smad2 Ser465/467 phosphorylation in TGFbeta-stimulated, but not unstimulated HepG2 cells. The corresponding target phosphoserine residues in the alternatively spliced human Smad2Deltaexon3 (Isoform Short) are pSer435 & pSer437 (UniProt Q15796-2).

Immunogen

Epitope: Phospho-Ser465/467.
KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Ser465/467 of human Smad2.

Application

Anti-phospho SMAD2 Antibody (Ser465/467) is an antibody against phospho SMAD2 for use in Western Blotting.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected Smad2 Ser465/467 phosphorylation in 10 µg lysate from TGFbeta-stimulated, but not unstimulated, HepG2 cells.

Target description

~53 kDa 52.31 kDa (Isoform Long) and 48.96 kDa (Isoform Short; Smad2Deltaexon3) calculated.

Linkage

Replaces: AB3849

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ksenia M Yegodayev et al.
Cancers, 12(2) (2020-02-08)
Most head and neck cancer (HNC) patients are resistant to cetuximab, an antibody against the epidermal growth factor receptor. Such therapy resistance is known to be mediated, in part, by stromal cells surrounding the tumor cells; however, the mechanisms underlying
Su Jung Oh-Hohenhorst et al.
Cancer gene therapy (2021-01-09)
The outcome of prostate cancer (PCa) patients is highly variable and depends on whether or not distant metastases occur. Multiple chromosomal deletions have been linked to early tumor marker PSA recurrence (biochemical relapse, BCR) after radical prostatectomy (RP), but their
Aditya Prakash Soni et al.
Frontiers in plant science, 13, 922694-922694 (2022-06-18)
The production of recombinant proteins in plant systems is receiving wider attention. Indeed, various plant-produced pharmaceuticals have been shown to be biologically active. However, the production of human growth factors and cytokines in heterologous systems is still challenging because they
Eoin D O'Sullivan et al.
JCI insight, 7(22) (2022-12-13)
Progressive fibrosis and maladaptive organ repair result in significant morbidity and millions of premature deaths annually. Senescent cells accumulate with aging and after injury and are implicated in organ fibrosis, but the mechanisms by which senescence influences repair are poorly
Shannon Romano et al.
Development (Cambridge, England), 147(18) (2020-09-09)
Sex determination and differentiation is a complex process regulated by multiple factors, including factors from the germline or surrounding somatic tissue. In zebrafish, sex-determination involves establishment of a bipotential ovary that undergoes sex-specific differentiation and maintenance to form the functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service