Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS445

Sigma-Aldrich

Anti-Caspase-5 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Caspase-5, CASP-5, ICE(rel)-III, Protease ICH-3, Protease TY, Caspase-5 subunit p202.Caspase-5 subunit p10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CASP5(838)

General description

Caspase-5 (CASP5) is a member of the peptidase C14A family that can be divided into the apoptotic and inflammatory caspase subfamilies. Members of this subfamily include caspase-1, -4, -5, and -12. When activated, these caspases result in the cleavage and activation of proinflammatory cytokines such as IL-1beta and IL-18. Caspase-5 is expressed in barely detectable amounts in most tissues except brain, with highest levels being found in lung, liver and skeletal muscle. Overexpression of the active form of this enzyme induces apoptosis in fibroblasts.

Specificity

This antibody recognizes the intermediate domain of Caspase-5.

Immunogen

Epitope: Intermediate domain
KLH-conjugated linear peptide corresponding to the intermediate domain of human Caspase-5.

Application

Detect Caspase 5 using this rabbit polyclonal antibody, Anti-Caspase-5 Antibody validated for use in western blotting, ICC & Immunofluorescence.
Immunocytochemistry Analysis: 2 µg/mL from a representative lot detected Caspase-5 in P815 cells.

Immunofluorescence Analysis: 2 µg/mL from a representative lot detected Caspase-5 in P815 cells.
Research Category
Signaling
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in Ramos cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Caspase-5 in Ramos cell lysate.

Target description

~48 kDa observed. Uncharacterized bands may appear in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Ramos cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service