Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS1634

Sigma-Aldrich

Anti-LIP5

serum, from rabbit

Synonym(s):

Vacuolar protein sorting-associated protein VTA1 homolog, Dopamine-responsive gene 1 protein, DRG-1, LYST-interacting protein 5, LIP5, SKD1-binding protein 1, SBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... VTA1(51534)

General description

Vacuolar protein sorting-associated protein VTA1 homolog (UniProt: Q9NP79; also known as Dopamine-responsive gene 1 protein; DRG-1; LYST-interacting protein 5; LIP5; SKD1-binding protein 1; SBP1) is encoded by the VTA1 (also known as C6orf55; HSPC228; My012) gene (Gene ID 51534) in human. LIP5 is a 33.9 kDa protein involved in trafficking of the multi-vesicular body. It helps regulate the membrane fission and recycling activities of the ESCRT pathway. It performs its central regulatory function within the ESCRT pathway by binding ESCRT-III proteins through its N-terminal tandem microtubule-interacting and trafficking (MIT) domain and by binding VPS4 ATPases through its C-terminal VSL (Vta1/SBP1/LIP5) domain. These interactions allow LIP5 to coordinate ESCRT-III and VPS4 activities both during normal cellular homeostasis and also in response to interferon signaling during innate immune responses. The ESCRT-III and VPS4 ATPase complexes catalyze the membrane fission events associated with these processes. The LIP5 protein helps regulate their interactions by binding directly to a subset of ESCRT-III proteins and to VPS4.

Specificity

Detects LIP5 (Vacuolar protein sorting-associated protein VTA1 homolog) in HeLa cells

Immunogen

His-tagged full length human recombinant LIP5.

Application

Detect LIP5 (Vacuolar protein sorting-associated protein VTA1 homolog), using this rabbit polyclonal Anti-LIP5, Cat. No. ABS1634, validated for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected LIP5 in HeLa cells.

Western Blotting Analysis: A representative lot detected endogenous LIP5 co-immunoprecipiated with Myc-tagged VPS4A from HEK293T cells (Skalicky, J.J., et al. (2012). J Biol Chem. 287(52):43910-43926).








Research Category
Signaling

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antiserum detected LIP5 in HeLa cell lysate.

Target description

~38 kDa observed. 33.88 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antibody serum with 0.05% sodium azide.
Unpurified.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service