Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABS314

Sigma-Aldrich

Anti-APPL1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DCC-interacting protein 13-alpha, Dip13-alpha, Adapter protein containing PH domain, PTB domain and leucine zipper motif 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
ICC
IHC
IP
WB
species reactivity:
rat, mouse, human
technique(s):
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, mouse, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... APPL1(26060)

General description

Adapter protein containing PH domain, PTB domain and leucine zipper motif 1 (APPL1) is also called DCC-interacting protein 13-alpha (Dip13-alpha). APPL1 is necessary for cell proliferation in response to extracellular signals and links Rab5 to nuclear signal transduction. APPL1 is expressed at high levels in heart, ovary, pancreas, and skeletal muscle tissues.

Immunogen

Recombinant protein corresponding to human APPL1.

Application

Detect Adapter protein containing PH domain, PTB domain & leucine zipper motif 1 using this rabbit polyclonal antibody, Anti-APPL1 Antibody validated for use in western blotting, IP, ICC & IHC.
Research Category
Apoptosis & Cancer
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blotting Analysis: 0.5 µg/mL of this antibody detected APPL1 in 10 µg of L6 and NIH/3T3 cell lysate.

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated APPL1 in HUVEC cell lysate (Lin, D. C., et al. (2006). Mol Cell Biol. 26(23):8928-8941.).

Immunohistochemistry Analysis: A representative lot from an independent laboratory detected APPL1 in liver tissues from mice fed with either a standard diet, high fat diet, or a high fat diet with exercise (Marinho, R., et al. (2012). J Cell Physiol. 227(7):2917-2926.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected APPL1 in pancreatic islets and impaired GSIS cells of dietary and obese mice (Cheng, K. K., et al. (2009). Cell Metab. 9(5):417-427.).

Quality

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected APPL1 in 10 µg of A431 cell lysate.

Target description

~84 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aparajita Lahree et al.
Cell reports, 39(9), 110886-110886 (2022-06-02)
Intracellular pathogens manipulate host cells to survive and thrive. Cellular sensing and signaling pathways are among the key host machineries deregulated to favor infection. In this study, we show that liver-stage Plasmodium parasites compete with the host to sequester a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service