Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5778

Sigma-Aldrich

Anti-Nurr1 / NOT1 Antibody

Chemicon®, from rabbit

Synonym(s):

Anti-HZF-3, Anti-NOT, Anti-NURR1, Anti-RNR1, Anti-TINUR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NR4A2(4929)

Specificity

Recognizes Nurr1 / NOT1 (Cys4 zinc finger of nuclear receptor type family), a zinc finger transcription factor.

Immunogen

Synthetic peptide.

Application

Detect Nurr1 / NOT1 using this Anti-Nurr1 / NOT1 Antibody validated for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Neurodegenerative Diseases
Western blot: 1:200-1:1,000 using ECL.

Immunohistochemistry: 1:200-1:1,000.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Precipitated antibody in a solution of 50% saturated ammonium sulfate and PBS containing no preservatives.

Storage and Stability

Maintain unopened vial at -70°C for up to 6 months. Avoid repeated freeze/thaw cycles.

PREPARATION AND USE:

To reconstitute the antibody, centrifuge the antibody vial at moderate speed (5,000 rpm) for 5 minutes to pellet the precipitated antibody product. Carefully remove the ammonium sulfate/PBS buffer solution and discard. It is not necessary to remove all of the ammonium sulfate/PBS solution: 10 μL of residual ammonium sulfate solution will not affect the resuspension of the antibody. Do not let the protein pellet dry, as severe loss of antibody reactivity can occur.

Resuspend the antibody pellet in a suitable biological buffer such as PBS or TBS (pH 7.3-7.5) to a final concentration of 1.0 mg/mL. For example, to achieve a 1.0 mg/mL concentration with 50 μg of precipitated antibody, the amount of buffer needed would be 50 μL.

Carefully add the liquid buffer to the pellet. DO NOT VORTEX. Mix by gentle stirring with a wide pipet tip or gentle finger-tapping. Let the precipitated antibody rehydrate for 1 hour at 4-25°C prior to use. Small particles of precipitated antibody that fail to resuspend are normal. Vials are overfilled to compensate for any losses.

The rehydrated antibody solutions can be stored undiluted at 2-8°C for 2 months without any significant loss of activity. Note, the solution is not sterile, thus care should be taken if product is stored at 2-8°C.

For storage at -20°C, the addition of an equal volume of glycerol can be used, however, it is recommended that ACS grade or higher glycerol be used, as significant loss of activity can occur if the glycerol used is not of high quality.

For long-term storage at -70°C, it is recommended that the rehydrated antibody solution be further diluted 1:1 with a 2% BSA (fraction V, highest-grade available) solution made with the rehydration buffer. The resulting 1% BSA/antibody solution can be aliquoted and stored frozen at -70°C for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl G Gottschalk et al.
Molecular neurobiology, 56(11), 7872-7887 (2019-05-28)
Nuclear receptor related 1 protein (Nurr1) is an important transcription factor required for differentiation and maintenance of midbrain dopaminergic (DA) neurons. Since decrease in Nurr1 function either due to diminished expression or rare mutation is associated with Parkinson's disease (PD)
Dopaminergic-like neurons derived from oral mucosa stem cells by developmental cues improve symptoms in the hemi-parkinsonian rat model.
Ganz, J; Arie, I; Buch, S; Zur, TB; Barhum, Y; Pour, S; Araidy, S; Pitaru, S; Offen, D
Testing null
Josh Cutts et al.
Methods in molecular biology (Clifton, N.J.), 1516, 121-144 (2016-04-24)
Neural progenitor cells (NPCs) derived from human pluripotent stem cells (hPSCs) are a multipotent cell population capable of long-term expansion and differentiation into a variety of neuronal subtypes. As such, NPCs have tremendous potential for disease modeling, drug screening, and
Theodora Papanikolaou et al.
The European journal of neuroscience, 29(12), 2264-2275 (2009-06-11)
The transcription factor Pitx3 is expressed exclusively by mesodiencephalic dopaminergic neurons; however, ablation of Pitx3 results in selective degeneration of primarily dopaminergic neurons of the substantia nigra pars compacta, the neuronal population that is most vulnerable in Parkinson's disease. Although
Kavina Ganapathy et al.
Journal of cellular biochemistry, 117(12), 2719-2736 (2016-04-12)
Post mortem studies on familial and sporadic Parkinson's disease patient striatal tissue have shown that nearly 90% of α-synuclein deposited in Lewy-bodies is phosphorylated at serine-129 (pSyn-129) as opposed to only 4% in normal human brain. We aimed to find

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service