Key Documents
Z256811
Ace double-jacketed coiled condenser with Ace-Thred hose connections
jacket L 250 mm
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 5956143
jacket L
250 mm
Related Categories
General description
With #7 Ace-Thred hose connections for 1/4 in. I.D. tubing. Extra cooling capacity for volatile materials. Features threaded inlet/outlet connections that incorporate UHDPE (ultra-high-density PE) connectors and nylon bushings to make easy connect/disconnect of coolant tubing . The bushing compresses the connector and a silicone O-ring for a leaktight seal. Compact design retards diffusion.
Legal Information
Ace-Thred is a trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)