Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

ABE2626M

Sigma-Aldrich

Anti-PASD1

from rabbit

Synonym(s):

Circadian clock protein PASD1, Cancer/testis antigen 63, CT63, OX-TES 11, PAS domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PASD1(139135)

General description

Circadian clock protein PASD1 (UniProt: Q8IV76; also known as Cancer/testis antigen 63, CT63, OC-TES 11, PAS domain-containing protein 1) is encoded by the PASD1 gene (Gene ID: 139135) in human. PASD1 is evolutionarily nuclear protein that associates preferentially at the periphery of the nucleus with heterochromatin. Human PASD1 contains an N-terminal PAS domain (a.a. 30-102), a central region (a.a. 365-412) essential for its transcriptional repression activity, and two coiled coil domains (CC1/a.a. 365-412 and CC2/a.a. 475-553), where CC1 exhibits significant homology with CLOCK exon 19-coded sequence. PASD1 is related to CLOCK and interacts with the bHLH-PAS transcription complex CLOCK-BMAL1 to repress transcriptional activation. PASD1 expression is normally limited to immunologically restricted germline tissues that do not express class I MHC molecules, such as testis and placenta, allowing tissue-specific circadian rhythms suppression. It is also expressed in a broad range of cancer cells, including melanoma, lung cancer, and breast cancer. It displays widespread expression in solid tumors and diffuse large B-cell lymphoma (DLBCL)-derived cell lines. Isoform 2 is expressed in all DLBCL-derived cell lines, while isoform 1 is preferentially expressed in cell lines derived from non-germinal center DLBCL. When over-expressed, PASD1 inhibits circadian clock function in cancer cells and reducing PASD1 in cancer cells significantly increases the amplitude of transcriptional oscillations to generate more robust circadian rhythms.

Specificity

This rabbit polyclonal antibody detects PASD1, isoform 1 version 2, in human cells. It targets an epitope within 19 amino acids from the C-terminal end.

Immunogen

A linear peptide corresponding to 19 amino acids from the C-terminal end of human PASD1, isoform 1.
Epitope: C-terminus

Application

Anti-PASD1, Cat. No. ABE2626, is a highly specific rabbit polyclonal antibody that targets Circadian clock protein PASD1 and has been tested in Immunoprecipitation and Western Blotting.
Research Category
Epigenetics & Nuclear Function
Western Blotting Analysis: A representative lot detected PASD1 in H1299 cells (Michael, A.K., et. al. (2015). Mol Cell. 58(5):743-54).

Immunoprecipitation Analysis: 5 ug from a representative lot immunoprecipitated PASD1 in 500 µg of SW480 lysate.

Western Blotting Analysis: A representative lot detected PASD1 in HEK293T transfected myc-his Tagged PASD1 lysate V2 isoform (Courtesy of Dr Carrie Partch at University of California Santa Cruz).

Immunoprecipitation Analysis: A representative lot detected PASD1 in H1299 & SW480 cells (Courtesy of Dr Carrie Partch at University of California Santa Cruz).

Immunoprecipitation Analysis: A representative lot detected PASD1 in H1299 and SW480 cell lysates (Michael, A.K., et. al. (2015). Mol Cell. 58(5):743-54).

Quality

Evaluated by Western Blotting in HEK293T cells transfected myc-His Tagged PASD1 lysate V2 isoform.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected PASD1 in lysate from HEK293T cells transfected myc-His Tagged PASD1, isoform 1, version2.

Target description

~120 kDa observed; 87.43 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service