Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABD101

Sigma-Aldrich

Anti-ESRRB Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Steroid hormone receptor ERR2, ERR beta-2, Estrogen receptor-like 2, Estrogen-related receptor beta, ERR-beta, Nuclear receptor subfamily 3 group B member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.75

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ESRRB(2103)

General description

The estrogen-related receptor beta (ESRRB) was initially identified in mouse embryonic carcinoma stem cells and was found to be expressed exclusively in trophoblast progenitor cells between days 6.5 and 7.5 post coitum. Recent studies have shown that ESRRB is part of an POU5F1/Oct4-centered protein interaction network in embryonic stem (ES) cells. It is thought to interact with other ES transcription factors such as NANOG within this network, activating POU5F1 transcription and thereby sustaining self-renewal and pluripotency in ES cells.

Immunogen

KLH-conjugated linear peptide corresponding to Amino acids 376-388 of Human ESRRB protein.

Application

Immunocytochemical Analysis: 1:50 dilution of this antibody detected ESRRB in a mouse embryonic stem cells.
This Anti-ESRRB Antibody is validated for use in Western Blotting and Immunocytochemistry for the detection of ESRRB.

Quality

Evaluated by Western Blot in a mouse brain E16 lysate. Western Blot Analysis: 1μg/ml of this antibody detected ESRRB in a mouse brain E16 lysate. Uncharacterized bands may be observed in some cell lysates with longer exposure times.

Target description

~56 kDa observed

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniela Cornacchia et al.
Cell stem cell, 25(1), 120-136 (2019-06-04)
Current challenges in capturing naive human pluripotent stem cells (hPSCs) suggest that the factors regulating human naive versus primed pluripotency remain incompletely defined. Here we demonstrate that the widely used Essential 8 minimal medium (E8) captures hPSCs at a naive-to-primed intermediate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service