Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC124

Sigma-Aldrich

Anti-phospho-ULK1 (Ser555) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Serine/threonine-protein kinase ULK1, Autophagy-related protein 1 homolog, ATG1, hATG1, Unc-51-like kinase 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), Xenopus (based on 100% sequence homology), mouse (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

inhibition assay: suitable (peptide)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer555)

Gene Information

human ... ULK1(8408)

General description

ULK1 or autophagy-related protein 1 homolog (ATG1) is a serine/threonine protein kinase. It is a ubiquitously expressed protein that is highly abundant in the brain, skeletal muscle, and heart. It is involved in the signaling pathway of mTOR receptors leading to the regulation of autophagy in nutrient-deprived cells. mTOR may interact with ULK1 as a complex of ULK1/FIP200/Atg13. ULK1 may play an important role in neuronal development: Elimination of ULK1′s enzymatic activity by targeted disruption of its kinase domain has blocked the outgrowth of neurons in the developing mouse brain, possibly by blocking the ULK1-mediated trafficking of neurotrophins.

Specificity

This antibody recognizes ULK1 phosphorylated at Ser555.

Immunogen

Epitope: Phosphorylated Ser555
KLH-conjugated linear peptide corresponding to human ULK1 phosphorylated at Ser555.

Application

Peptide Inhibition Analysis: A representative lot of this antibody blocked ULK1 in PIA involving peptide 3123-C vs. peptide 3123 on serum starved MCF7 cell lysates.
Research Category
Apoptosis & Cancer
Research Sub Category
Kinases & Phosphatases
This Anti-phospho-ULK1 (Ser555) Antibody is validated for use in Western Blotting, Peptide Inhibition Assay for the detection of phospho-ULK1 (Ser555).

Quality

Evaluated by Western Blot in non-serum starved and serum starved MCF7 cell lysates.

Western Blot Analysis: 0.2 µg/mL of this antibody detected phospho-ULK1 (Ser555) in 10 µg of non-serum starved and serum starved MCF7 cell lysates.

Target description

~120 kDa observed. Uniprot describes a molecular weight at 112 kDa This protein has been observed at greater than ~125 kDa due to multiple phosphorylation-dephosphorylation processes. (Ganley, I. G., et al. (2009). J Biol Chem. 284(18):12297-12305.)

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Non-serum starved and serum starved MCF7 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PHY906(KD018), an adjuvant based on a 1800-year-old Chinese medicine, enhanced the anti-tumor activity of Sorafenib by changing the tumor microenvironment.
Lam, W; Jiang, Z; Guan, F; Huang, X; Hu, R; Wang, J; Bussom, S; Liu, SH; Zhao, H; Yen, Y; Cheng, YC
Scientific Reports null
Aleksandra V Borodkina et al.
Cell cycle (Georgetown, Tex.), 15(1), 117-127 (2015-12-05)
Previously we demonstrated that endometrium-derived human mesenchymal stem cells (hMESCs) via activation of the ATM/p53/p21/Rb pathway enter the premature senescence in response to oxidative stress. Down regulation effects of the key components of this signaling pathway, particularly ATM and p53
Ryan N Montalvo et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 318(2), R227-R233 (2019-11-28)
Doxorubicin (DOX) is a highly effective antineoplastic agent used in cancer treatment. Unfortunately, clinical use of DOX is limited due to the development of dose-dependent toxicity to cardiac and respiratory (i.e., diaphragm) muscles. After administration, DOX preferentially localizes to the
Insu Kwon et al.
The journal of physiological sciences : JPS, 68(3), 269-280 (2017-02-19)
Elevation of anabolism and concurrent suppression of catabolism are critical metabolic adaptations for muscular hypertrophy in response to resistance exercise (RE). Here, we investigated if RE-induced muscular hypertrophy is acquired by modulating a critical catabolic process autophagy. Male Wistar Hannover
Aleksandra V Borodkina et al.
Aging, 8(12), 3400-3418 (2016-12-13)
Intracellular calcium ([Ca2+]i) has been reported to play an important role in autophagy, apoptosis and necrosis, however, a little is known about its impact in senescence. Here we investigated [Ca2+]i contribution to oxidative stress-induced senescence of human endometrium-derived stem cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service