Skip to Content
MilliporeSigma
All Photos(4)

Documents

78300BC

Millipore

MVP ICON® Instrument

BioControl, ICON ATP Monitoring for HACCP Management

Synonym(s):

ATP Analyzer, HACCP management system, MVP ICON® Device, Quality Management Instrument, Reliable Quality Control Equipment

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115333

Agency

EP
ISO
USP

product line

MVP ICON®

feature

ready-to-use

packaging

pkg of 1 ea

application(s)

food and beverages

detection method

bioluminescent

shipped in

ambient

General description

The MVP ICON® is a complete pioneering HACCP and hygiene monitoring system. The system measures and records Adenosine Triphosphate (ATP) and other key HACCP parameters (pH, temperature, conductivity, and chemical concentration) to provide a management tool that ensures the success of the quality assurance program. It can measure ATP in seconds. The software displays the results on a proprietary Zones of Cleanliness scale that has been adapted from RLU′s. The results are categorized into clean, cautious, and fail areas, facilitating prompt identification and implementation of necessary corrective actions.The MVP ICON® instrument comes with a multi-variable platform and responsive touch display. It also facilitates on-site calibration for all parameters. The MVP ICON® Dashboard software offers a live overview of critical program metrics vital for effective quality management of the HACCP program. The design of the sampling devices compensates for variations in the swabbing technique, ensuring accurate and consistent readings. The proprietary reagents prevent chemical interference, allowing for ATP testing before or after sanitizing. The MVP ICON® system Conductivity Probe allows precise measurement of chemical concentrations to avoid unnecessary over-use of expensive chemicals. Probes for pH and temperature are also available.

Application

MVP ICON® is designed for rapid monitoring of surface hygiene and cleaning efficacy indicators (such as ATP (Adenosine Triphosphate), NAD (Nicotinamide Adenine Dinucleotide), and protein detection). It is a comprehensive hygiene monitoring system for the Food and Beverage industries.

Features and Benefits

  • Superior ATP swab design with best-in-class sample release
  • Ready-to-use room-temperatures table swabs
  • Flexible testing options that complement existing workflows
  • Perform instrument calibrations and verifications onsite, anytime.
  • Easily track failure rates, retest compliance, and monitor overall program effectiveness

Components

  • MVP ICON Instrument
  • Protective Case
  • USB Cable
  • AC Adapter
  • International Adapters
  • Shoulder Strap
  • Quick Start Guide

Suitability

ATP Testing, Food Processing, HACCP Monitoring, Sanitation

Principle

The detection of ATP through bioluminescence is a well-established and widely used indicator of surface hygiene and cleaning efficacy in food processing. Visual inspection is not an adequate critical control for surface hygiene and the lengthy time-to-result of microbiological test methods (from 2 - 7 days) make them ineffective for hygiene monitoring. ATP detection with the MVP ICON provides results in 10 seconds allowing for immediate "go/no-go" decisions to be made prior to the start of processing and allows for correction actions, when necessary, to be taken immediately. ATP is a molecule that is found in all living cells, including: bacteria, yeast, mold and all organic matter. After cleaning and/or sanitizing, food processing equipment and the plant environment is tested for ATP. If cleaning/sanitizing has been effective, ATP levels should be low.

Testing Method

ATP Bioluminescence

System Requirements

  • Windows 7 or Windows 10
  • Minimum memory requirement 512 MB; recommend at least 1GB or more of RAM
  • At least 16 GB of hard disk space
  • WXGA video resolution of 1280 x 800 or better
  • Mouse or other pointing device
  • One available USB port
  • Adobe Acrobat Reader or similar PDF Viewer
  • Printer (optional)

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
MVP ICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Media fill tests for beverages ensure that the filling line is not contaminated with spoilage bacteria, yeasts, or molds, during the production of low-acid aseptic beverages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service