Skip to Content
MilliporeSigma
All Photos(3)

Documents

AB9864R

Sigma-Aldrich

Anti-NMDAR1 Antibody, clone 1.17.2.6, rabbit monoclonal

clone 1.17.2.6, from rabbit, unconjugated

Synonym(s):

N-methyl-D-aspartate receptor channel, subunit zeta-1, N-methyl-D-aspartate receptor subunit NR1, NMDA receptor 1, glutamate [NMDA] receptor subunit zeta 1, glutamate receptor, ionotropic, N-methyl D-aspartate 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1.17.2.6, monoclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRIN1(2902)
mouse ... Grin1(14810)
rat ... Grin1(24408)

General description

N-methyl-D-aspartate receptor subunit NR1 (NMDAR1) is a multi-pass membrane protein belonging to the glutamate-gated ion channel family and is one of two subunits comprising the tetrameric protein NMDAR localized throughout the brain at excitatory synapses. NMDAR1 is mediated by glycine and is a critical component to excitotoxicity, learning, memory acquisition, synaptic plasticity, and synaptogenesis. It also plays a regulatory role for neuronal functions involving glutamate neurotransmission and the targeting of NMDA receptors on the cell surface. As a glutamate-gated ion channel, NMDAR1 is highly calcium permeable and possesses sensitivity to magnesium in a voltage-dependent manner.

Immunogen

Epitope: Recognizes NMDAR1 at the C-terminus
Linear peptide corresponding to human NMDAR1.

Application

Anti-NMDAR1 Antibody, clone 1.17.2.6 detects level of NMDAR1 & has been published & validated for use in WB, IH.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western Blot Analysis: 0.1 µg/mL of this antibody from a previous lot detected NMDAR1 on 10 µg of mouse microsomal brain tissue lysate.

Immunohistochemistry Analysis: 1:250 dilution of this antibody from a previous lot detected NMDAR1 in rat cerebellum tissue.

Quality

Evaluated by Western Blot in rat brain tissue lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected NMDAR1 on 10 µg of rat brain tissue lysate.

Target description

~120 kDa observed

Physical form

Protein A
Purified rabbit monoclonal IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide and 40% Glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Rat brain tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Milena De Felice et al.
Molecular pain, 12 (2016-04-21)
Bone metastases occur frequently in advanced breast, lung, and prostate cancer, with approximately 70% of patients affected. Pain is a major symptom of bone metastases, and current treatments may be inadequate or have unacceptable side effects. The mechanisms that drive
Mirko Feldmann et al.
Cerebral cortex (New York, N.Y. : 1991), 29(2), 892-905 (2018-12-12)
Although by adulthood cortical structures and their capacity for processing sensory information have become established and stabilized, under conditions of cortical injury, or sensory deprivation, rapid reorganization occurs. Little is known as to the impact of this kind of adaptation
Juan Hong et al.
Neural plasticity, 2018, 3989201-3989201 (2018-08-21)
Cochlear inner hair cell (IHC) ribbon synapses play an important role in sound encoding and neurotransmitter release. Previous reports show that both noise and aminoglycoside exposures lead to reduced numbers and morphologic changes of synaptic ribbons. In this work, we
Mikel Pérez-Rodríguez et al.
Cerebral cortex (New York, N.Y. : 1991), 29(8), 3266-3281 (2018-09-01)
Critical periods of synaptic plasticity facilitate the reordering and refining of neural connections during development, allowing the definitive synaptic circuits responsible for correct adult physiology to be established. Presynaptic spike timing-dependent long-term depression (t-LTD) exists in the hippocampus, which depends
Yasuhito Watanabe et al.
Frontiers in molecular neuroscience, 8, 87-87 (2016-01-23)
N-methyl-D-aspartate receptors (NMDARs) in all hippocampal areas play an essential role in distinct processes of memory formation as well as in sustaining cell survival of postnatally generated neurons in the dentate gyrus (DG). In contrast to the beneficial effects, over-activation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service