Skip to Content
MilliporeSigma
All Photos(1)

Documents

MAB1586

Sigma-Aldrich

Anti-NMDAR1 Antibody, (all splice variants), clone R1JHL

clone R1JHL, Chemicon®, from mouse

Synonym(s):

Anti-GluN1, Anti-NDHMSD, Anti-NDHMSR, Anti-NMD-R1, Anti-NMDA1, Anti-NMDAR1, Anti-NR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

R1JHL, monoclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

Not specified

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GRIN1(2902)
mouse ... Grin1(14810)
rat ... Grin1(24408)

Specificity

NMDA receptor subunit 1 (all splice variants). No cross-reactivity observed with other NMDA receptor subunits. Recognizes a 103-116 kDa protein in rat brain hippocampal homogenate by immunoblot.

Immunogen

Epitope: (all splice variants)
Fusion protein of amino acids 1-564 from rat NR1.

Application

Detect NMDAR1 using this Anti-NMDAR1 Antibody, (all splice variants), clone R1JHL validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1:300. It is recommended that the antibody be diluted in TBST (20 mM Tris-HCl, 137 mM NaCl, 1% Tween-20, pH 7.6) containing 5% nonfat milk and 0.05% sodium azide. The diluted antibody can be used at least 5 times without a significant signal decrease. Store at 2-8°C between uses.

Immunoprecipitation: 3 μg per 200 μg of lysate will quantitatively precipitate all the NMDA R1 in 200 μg of rat brain lysate.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin. Lyophilized. Reconstitute with 50 μL of PBS, pH 7.4. Contains no preservative.
Format: Purified

Storage and Stability

Maintain lyophilized material at -20°C for up to six months after date of receipt. After reconstitution, maintain at -20°C in undiluted aliquots for up to six months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lenka Mikasova et al.
Brain : a journal of neurology, 135(Pt 5), 1606-1621 (2012-05-01)
Autoimmune synaptic encephalitides are recently described human brain diseases leading to psychiatric and neurological syndromes through inappropriate brain-autoantibody interactions. The most frequent synaptic autoimmune encephalitis is associated with autoantibodies against extracellular domains of the glutamatergic N-methyl-d-aspartate receptor, with patients developing
Meilin Tian et al.
Scientific reports, 6, 34751-34751 (2016-10-08)
Allostery is essential to neuronal receptor function, but its transient nature poses a challenge for characterization. The N-terminal domains (NTDs) distinct from ligand binding domains are a major locus for allosteric regulation of NMDA receptors (NMDARs), where different modulatory binding
María Rodríguez-Muñoz et al.
Oncotarget, 6(34), 35458-35477 (2015-10-16)
In the brain, the histidine triad nucleotide-binding protein 1 (HINT1) and sigma 1 receptors (σ1Rs) coordinate the activity of certain G-protein coupled receptors (GPCRs) with that of glutamate N-methyl-D-aspartate receptors (NMDARs). To determine the role of HINT1-σ1R in the plasticity
Nami Tajima et al.
Nature, 534(7605), 63-68 (2016-05-03)
The physiology of N-methyl-d-aspartate (NMDA) receptors is fundamental to brain development and function. NMDA receptors are ionotropic glutamate receptors that function as heterotetramers composed mainly of GluN1 and GluN2 subunits. Activation of NMDA receptors requires binding of neurotransmitter agonists to
Katharine R Smith et al.
The Journal of biological chemistry, 292(49), 20173-20183 (2017-10-20)
Brain ischemia causes oxygen and glucose deprivation (OGD) in neurons, triggering a cascade of events leading to synaptic accumulation of glutamate. Excessive activation of glutamate receptors causes excitotoxicity and delayed cell death in vulnerable neurons. Following global cerebral ischemia, hippocampal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service