Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB6001

Sigma-Aldrich

Anti-MMP-9 Antibody, NT

from rabbit, purified by affinity chromatography

Synonym(s):

92 kDa gelatinase, 92 kDa type IV collagenase, Gelatinase B, macrophage gelatinase, matrix metallopeptidase 9, matrix metalloproteinase 9, type V collagenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

canine, human

species reactivity (predicted by homology)

primate (based on 100% sequence homology), pig (based on 100% sequence homology), dog (based on 100% sequence homology), rat (based on 100% sequence homology), horse (based on 100% sequence homology), monkey (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MMP9(4318)

General description

All cells within tissues are surrounded by an extracellular matrix (ECM) giving the tissues shape and structure. The ECM is constantly being remodeled and constant communication is maintained between cells through this matrix. Secreted proteins, termed matrix metalloproteinases (MMPs), are involved in the modulation of cell matrix interactions. MMPs are Zn2+ binding endopeptidases that degrade various components of the ECM. MMPs are enzymes implicated in normal and pathologic tissue remodeling processes, wound healing, angiogenesis, and tumor invasion. These enzymes are very potent when active, and are associated with extracellular space inhibitors called TIMPs (tissue inhibitors of matrix metalloproteinases).

MMP9, also known as gelatinase B, is a secreted enzyme which degrades the interstitial collagens, types I, II, and III and is produced by normal alveolar macrophages and granulocytes.

Specificity

The antibody recognizes human MMP-9. It does not cross react with MMP-1 or MMP-2.

Immunogen

Epitope: N-terminus
KLH-conjugated synthetic peptide corresponding to the N-terminus of MMP-9.

Application

Anti-MMP-9 Antibody, N-terminus is an antibody against MMP-9 for use in WB.
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs

Quality

Evaluated on a representative lot by Western blot using an HL-60 cell lysate supplemented with a PMA conditioned media.

Western Blot Analysis: 0.5 µg/ml of this antibody detected MMP-9 on 10 µg of HL-60 plus PMA conditioned media.

Target description

92kDa (pro-form) and 88kDa (active form)

Linkage

Replaces: AB805

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HL-60 plus PMA conditioned media

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haiying Yue et al.
Experimental and therapeutic medicine, 10(4), 1437-1444 (2015-12-02)
Radiation-induced lung injury (RILI) is a common complication associated with thoracic radiotherapy. The aim of the present study was to investigate the effects of a single 15-Gy dose of right-thoracic lung irradiation on the expression levels of matrix metalloproteinases (MMPs)
Laura Mosca et al.
International journal of oncology, 56(5), 1212-1224 (2020-04-23)
S‑Adenosyl‑L‑methionine (AdoMet) is the principal methyl donor in transmethylation reactions fundamental to sustaining epigenetic modifications. Over the past decade, AdoMet has been extensively investigated for its anti‑proliferative, pro‑apoptotic and anti‑metastatic roles in several types of human cancer. Head and neck
Alexander C W Smith et al.
Nature neuroscience, 17(12), 1655-1657 (2014-10-20)
Relapse to cocaine use necessitates remodeling excitatory synapses in the nucleus accumbens and synaptic reorganization requires matrix metalloproteinase (MMP) degradation of the extracellular matrix proteins. We found enduring increases in MMP-2 activity in rats after withdrawal from self-administered cocaine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service