Skip to Content
MilliporeSigma

AB5467

Anti-AANAT Antibody

serum, Chemicon®

Synonym(s):

Arylalkylamine N-acetyltransferase, SNAT, Serotonin Acetylase, EC 2.3.1.87

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, sheep, fish, human, bovine

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AANAT(15)

Immunogen

Synthetic peptide (phosphorylated) corresponding to amino acids near the N-terminous of rat AANAT conjugated to BSA. {Ganguly, S et al, 2005}.

Application

Anti-AANAT Antibody is an antibody against AANAT for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western Blot: 1:10,000-1:20,000 using ECL. Reacts with the ~23 kDa AANAT protein. Some reactivity may also be seen at ~97 kDa. Suggested blocking buffer is PBS with 10% non fat dry milk and 0.2% Tween-20. Suggested dilution buffer is PBS with 1 mg/mL BSA fraction V. Suggested incubation time is overnight at room temperature (~23°C).

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Recognizes AANAT (Arylalkylamine N-acetyltransferase). The antibody specifically recognizes the phosphorylated N-terminal PKA site of AANAT (pT31) (Ganguly et al. 2005). The antibody reacts with natural and recombinant protein.

Physical form

Liquid rabbit serum containing no preservative.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL:

Night adapted rat pineal gland. Expression is light sensitive; highest levels are obtained from animals at night.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
AB546704053252330537