Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

AB5421-II

Sigma-Aldrich

Anti-Vesicular Glutamate Transporter 3 (VGLUT3)

from rabbit, purified by affinity chromatography

Synonym(s):

Solute carrier family 17 member 8, VGluT3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

Gene Information

General description

Vesicular glutamate transporter 3 (UniProt Q7TSF2; also known as Solute carrier family 17 member 8, VGluT3) is encoded by the Slc17a8 (also known as Vglut3) gene (Gene ID 266767) in rat species. VGluTs are proton-dependent carriers that are responsible for the upload of the major excitatory neurotransmitter glutamate into synaptic vesicles within presynaptic terminals to allow later regulated release at the synaptic cleft. VGluT3 is a multi-pass synaptic vesicle membrane protein that mediates the uptake of glutamate into synaptic vesicles at presynaptic nerve terminals of excitatory neurons. Rat VGluT3 has 12 helical, 7 cytoplasmic, and 6 vesicular domains. It is structurally similar to VGluT1 and -2, but differs in its distribution pattern. Unlike VGluT1 and 2 that rely on interactions with the endocytic machinery for recycling, but remain almost exclusively axonal, VGluT3 can target to the cell body and dendrites, as well as the axon of certain neurons and is implicated in the retrograde synaptic release of glutamate. VGluT3 displays a very restricted localization and is not present in known glutamatergic neurons, but is found in cholinergic neurons in the caudate-putamen and in serotonergic neurons in the raphe nucleus. VGluT3 is localized over vesicle clusters in both symmetric (inhibitory or modulatory) and asymmetric (excitatory) synapses. (Ref.: Herzog, E et al. (2004). Neuroscience. 123(4):983-1002; Seal, RP et al. (2008). Neuron 57(2); 263-275).

Specificity

This rabbit polyclonal antibody specifically detects Vesicular glutamate transporter 3(VGluT3). It targets an epitope with in 13 amino acids from the C-terminal region.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of rat Vesicular glutamate transporter 3 (VGluT3).

Application

Anti-Vesicular Glutamate Transporter 3 (VGLUT3), Cat. No. AB5421-II, is a highly specific rabbit polyclonal antibody that targets Vesicular glutamate transporter 3 and has been tested foe use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Research Category
Neuroscience
Western Blotting Analysis: A 1:500 dilution from a representative lot detected Vesicular Glutamate Transporter 3 (VGLUT3) in PC12 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected Vesicular Glutamate Transporter 3 (VGLUT3) in rat brain and human brain tissue sections.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Vesicular Glutamate Transporter 3 (VGLUT3) in PC12 cells.

Quality

Evaluated by Western Blotting in PC12 tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Vesicular Glutamate Transporter 3 (VGLUT3) in PC12 cell lysate.

Target description

~ 60 kDa observed; 64.76 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service