Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1786P

Sigma-Aldrich

Anti-Dopamine D3 Receptor Antibody, cytoplasmic domain

Chemicon®, from rabbit

Synonym(s):

D3R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Drd3(13490)
rat ... Drd3(29238)

Specificity

Specific for Dopamine Receptor D3. Rat Dopamine D3 receptor is a 446 aa, G-protein coupled and transmembrane receptor protein (Giros et al. 1990, 1991; Schmauss et al. 1993; Liu et al. 1994; Fu et al. 1995; Sokoloff et al. 1990; Livingstone et al. 1992). The immunogen peptide shows no significant homology with other dopamine receptors (D1, D2, D4 or D5).

SPECIES REACTIVITIES: The immunogen peptide shows sequence homology of 88% with mouse and 63% with human/monkey (12/19 aa) D3 receptors (Giros et al. 1990, 1991; Schmauss et al. 1993; Liu et al. 1994; Fu et al. 1995; Sokoloff et al. 1990; Livingstone et al. 1992). Antibody cross-reactivity with various species is not known.

Immunogen

A 19 amino acid peptide sequence from the rat D3 receptor within the 3rd cytoplasmic domain.
Epitope: cytoplasmic domain

Application

Anti-Dopamine D3 Receptor Antibody, cytoplasmic domain detects level of Dopamine D3 Receptor & has been published & validated for use in ELISA, IH & WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western blot: 1-10 μg/mL (Chemiluminescence technique; 45-50 kDa) Approximately 100-400 μg of membrane protein/lane should be loaded to visualize the receptor band. In some systems 70kDa bands may also be present.

Immunohistochemistry: suggested working concentration is 2-10 μg/mL. Suggested fixative is 4% PFA, fixation 10-15′, permeabilized with 0.3-0.5% triton X-100 for 10-15′ (tissue, less time for cels), primary antibody dilution is 1:100 for direct fluorescence . {Coronas, V, et al. J Neurochemistry 91:1292-1301, 2004},

ELISA: 0.5-1.0 μg/mL (1 μg/mL immunogen peptide (Catalog Number AG229)/well)

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moderate Alcohol Exposure during the Rat Equivalent to the Third Trimester of Human Pregnancy Alters Regulation of GABAA Receptor-Mediated Synaptic Transmission by Dopamine in the Basolateral Amygdala.
Diaz, MR; Jotty, K; Locke, JL; Jones, SR; Valenzuela, CF
Frontiers in pediatrics null
Vito S Hernández et al.
Neuropharmacology, 82, 88-100 (2013-12-03)
L-Dopa is the major symptomatic therapy for Parkinson's disease, which commonly occurs in elderly patients. However, the effects of chronic use on mood and cognition in old subjects remain elusive. In order to compare the effects of a chronic pulsatile
K L Conrad et al.
Neuroscience, 169(1), 182-194 (2010-05-04)
Dopamine receptors (DARs) in the nucleus accumbens (NAc) are critical for cocaine's actions but the nature of adaptations in DAR function after repeated cocaine exposure remains controversial. This may be due in part to the fact that different methods used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service