Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1526

Sigma-Aldrich

Anti-Nerve Growth Factor-β Antibody

serum, Chemicon®

Synonym(s):

NGFbeta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat, chicken, human

should not react with

bovine

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NGF(4803)

Specificity

Nerve growth factor-beta (NGF). Less than 1% cross-reactivity against recombinant human NT3, NT4 and BDNF by ELISA.

Immunogen

Mouse nerve growth factor-beta purified from salivary glands.

Application

Immunohistochemistry at 1:500-1:5,000 (see suggested protocol).

Immunoblotting at 1:500-1:5,000.

ELISA at 1:500-1:5,000.

Inhibition of biological activity in vitro at 1:10-1:50.
BIOLOGICAL ACTIVITY: This antibody will completely inhibit the survival and neurite outgrowth actions of mouse NGF in chicken dorsal root ganglion neurons in vitro.

Use neat for in vivo animal studies.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins
This Anti-Nerve Growth Factor-β Antibody is validated for use in ELISA, WB, IH, FUNC for the detection of Nerve Growth Factor-β.

Linkage

Replaces: 04-1119

Physical form

Whole rabbit antisera. Lyophilized. Reconstitute with 250 μL of sterile distilled water.

Storage and Stability

Maintain at -20°C to -70°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsia-Shu Lo et al.
The journal of obstetrics and gynaecology research, 43(1), 173-178 (2016-10-21)
By investigating the association of urodynamics and urogenital nerve growth factor (NGF) levels in vaginal mesh surgery, we may be able to associate the likelihood of postoperative lower urinary tract symptoms developing as a result of synthetic mesh implanted for
Sashwati Roy et al.
Nanomedicine : nanotechnology, biology, and medicine, 28, 102220-102220 (2020-05-19)
This work rests on our recent report on the successful use of tissue nanotransfection (TNT) delivery of Ascl1, Brn2, and Myt1l (TNTABM) to directly convert skin fibroblasts into electrophysiologically active induced neuronal cells (iN) in vivo. Here we report that
Daniel Aridgides et al.
PloS one, 8(2), e57450-e57450 (2013-02-26)
Cardiomyocytes express neurotrophin receptor TrkA that promotes survival following nerve growth factor (NGF) ligation. Whether TrkA also resides in cardiac fibroblasts (CFs) and underlies cardioprotection is unknown. To test whether CFs express TrkA that conveys paracrine signals to neighbor cardiomyocytes
A Vega et al.
Developmental neurobiology, 76(8), 832-846 (2015-11-13)
Neurons are able to segregate transmitters to different axon endings. Segregation is a plastic neuronal feature; it can be modulated by synaptic environment. We have demonstrated that neurotrophin and other cellular factors regulate segregation in sympathetic neurons in culture. Herein
Caterina Porcino et al.
International journal of molecular sciences, 25(5) (2024-03-13)
Specific subpopulations of neurons in nerve and sensory systems must be developed and maintained, and this is accomplished in significant part by neurotrophins (NTs) and the signaling receptors on which they act, called tyrosine protein kinase receptors (Trks). The neurotrophins-tyrosine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service