Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

902885

Supelco

Cabinet wall duct 55 mm

10 mm connector (connections on both sides)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121815
NACRES:
NB.51

fitting

10 mm connector (connections on both sides)

shipped in

ambient

storage temp.

no temp limit

Related Categories

General description

Individual solvent supply installations often work with safety cabinets and and fume hoods. This cabinet wall duct can be used as the link between the inner and outer side of the safety cabinet with a max. wall thickness of 55 mm. Connection ends are 10 mm tube ends.
with 10 mm tubing connections on both sides, 22 mm diameter, 115 mm length

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service