Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.52085

Sigma-Aldrich

Fmoc-Glu(O-2-PhiPr)-OH

≥97% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-Glu(O-2-PhiPr)-OH, N-α-Fmoc-L-glutamic acid γ-2-phenylisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H29NO6
CAS Number:
Molecular Weight:
487.54
UNSPSC Code:
12352209
NACRES:
NA.22

product name

Fmoc-Glu(O-2-PhiPr)-OH, Novabiochem®

Quality Level

product line

Novabiochem®

Assay

≥90.0% (acidimetric)
≥95.0% (HPLC)
≥97% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

15-25°C

InChI

1S/C29H29NO6/c1-29(2,19-10-4-3-5-11-19)36-26(31)17-16-25(27(32)33)30-28(34)35-18-24-22-14-8-6-12-20(22)21-13-7-9-15-23(21)24/h3-15,24-25H,16-18H2,1-2H3,(H,30,34)(H,32,33)/t25-/m0/s1

InChI key

NPNRKQDJIWMHNG-VWLOTQADSA-N

General description

Quasi-orthogonally-protected Glu derivative. The 2-PhiPr group can be removed selectively in the presence of tBu-based protecting groups by treatment with 1% TFA in DCM [1], making this derivative an extremely useful tool for the preparation of cyclic peptides by Fmoc SPPS [2] or for library synthesis. For the on-resin synthesis of side-chain to side-chain lactam bridged peptides, the combination of Lys(Mtt) and Glu(O-2-PhiPr) is particularly advantageous since both side-chains can be simultaneously unmasked in a single step.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] C. Yue, et al. (1993) Tetrahedron Lett., 34, 323.
[2] F. Dick, et al. in ′Peptides 1996, Proc. of 24th European Peptide Symposium′, R. Ramage & R. Epton (Eds), Mayflower Scientific Ltd., Birmingham, 1998, pp. 339.

Linkage

Replaces: 04-12-1199

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(011A)): ≥ 97 %
Purity (TLC(0811)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 2.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service