Skip to Content
MilliporeSigma
All Photos(1)

Documents

8.52072

Sigma-Aldrich

Fmoc-Asp-OAll

Novabiochem®

Synonym(s):

Fmoc-Asp-OAll, N-α-Fmoc-L-aspartic acid α-allyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H21NO6
CAS Number:
Molecular Weight:
395.41
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

Assay

≥97.0% (acidimetric)
≥98% (TLC)
≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

95 °C (decomposes)

application(s)

peptide synthesis

functional group

carboxylic acid

storage temp.

2-30°C

InChI

1S/C22H21NO6/c1-2-11-28-21(26)19(12-20(24)25)23-22(27)29-13-18-16-9-5-3-7-14(16)15-8-4-6-10-17(15)18/h2-10,18-19H,1,11-13H2,(H,23,27)(H,24,25)/t19-/m0/s1

InChI key

ZJMVIWUCCRKNHY-IBGZPJMESA-N

General description

Orthogonally-protected building block for the synthesis of head-to-tail cyclic peptides [1,2,3,4,5] by Fmoc SPPS. The α-allyl ester can be selectively removed in the presence of Fmoc- and tBu-based protecting groups by treatment with Pd(Ph3P)4/ CHCl3/AcOH/NMM [6], thereby facilitating the synthesis of branched esters, amides, lactones and lactams incorporating an aspartyl unit.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Literature references

[1] W. Bannwarth, et al. (1992) Tetrahedron Lett., 33, 4557.
[2] F. Albericio, et al. (1993) Tetrahedron Lett., 34, 1549.
[3] J. Eichler, et al. (1994) Pept. Res., 7, 300.
[4] J. Eichler, et al. in ′Peptides 1994, Proc. 23rd European Peptide Symposium′, H. Maia (Eds), ESCOM, Leiden, 1994, pp. 461.
[5] J. Eichler, et al. in ′Solid Phase Synthesis & Combinatorial Libraries, 4th International Symposium′, R. Epton (Eds), Mayflower Scientific Ltd., Birmingham, 1996, pp. 201.
[6] S. A. Kates, et al. in ′Peptides, Chemistry, Structure & Biology, Proc. 13th American Peptide Symposium′, ESCOM, Leiden, 1994, pp. 113.

Linkage

Replaces: 04-12-1157

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): conforms
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(CMA2)): ≥ 98 %
Purity (TLC(157A)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 97.0 %
Water (K. F.): ≤ 1.5 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service