Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

439908

Sigma-Aldrich

1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea

98%

Synonym(s):

N,N′-Di-Boc-S-methylisothiourea, 1,3-Di-Boc-2-methylisothiourea, N,N′-Bis(tert-butoxycarbonyl)-S-methylisothiourea

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CN=C(SCH3)NHCO2C(CH3)3
CAS Number:
Molecular Weight:
290.38
Beilstein:
5815984
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

115-121 °C (lit.)

application(s)

peptide synthesis

SMILES string

CS\C(NC(=O)OC(C)(C)C)=N/C(=O)OC(C)(C)C

InChI

1S/C12H22N2O4S/c1-11(2,3)17-9(15)13-8(19-7)14-10(16)18-12(4,5)6/h1-7H3,(H,13,14,15,16)

InChI key

UQJXXWHAJKRDKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Liam Martin et al.
Biomacromolecules, 19(8), 3190-3200 (2018-06-12)
A range of well-defined guanidinium-rich linear polymers with demonstrable efficiency for cellular internalization were developed. A protected guanidinium-functional acrylamide monomer (di-Boc-guanidinium ethyl acrylamide, GEAdiBoc) was synthesized and then polymerized via RAFT polymerization to yield well-defined homopolymers, which were then deprotected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service