Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41296

Sigma-Aldrich

2-Nitrobenzyl chloride

for synthesis

Synonym(s):

2-Nitrobenzyl chloride, α-Chloro-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6ClNO2
CAS Number:
Molecular Weight:
171.58
MDL number:
UNSPSC Code:
12352101
EC Index Number:
210-300-5
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

powder

bp

127-133 °C/13 hPa

mp

44-48 °C

transition temp

flash point 112 °C

storage temp.

2-30°C

SMILES string

ClCc1c(cccc1)[N+](=O)[O-]

InChI

1S/C7H6ClNO2/c8-5-6-3-1-2-4-7(6)9(10)11/h1-4H,5H2

InChI key

BXCBUWKTXLWPSB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 44 °C
Melting range (upper value): ≤ 48 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F

Flash Point(C)

112 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service