Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41032

Sigma-Aldrich

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetracetic acid monohydrate

for synthesis

Synonym(s):

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetracetic acid monohydrate, 1,2-Cyclohexanediamine-N,N,N′N′-tetraacetic acid, 1,2-Cyclohexylenedinitrilotetraacetic acid, 1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O8 · xH2O
CAS Number:
Molecular Weight:
346.33 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
EC Index Number:
236-308-9
NACRES:
NA.22

Quality Level

form

powder

mp

213 °C

bulk density

500 kg/m3

storage temp.

2-30°C

SMILES string

N([C@H]1[C@@H](CCCC1)N(CC(=O)[O-])CC(=O)O)(CC(=O)[O-])CC(=O)O

InChI

1S/C14H22N2O8/c17-11(18)5-15(6-12(19)20)9-3-1-2-4-10(9)16(7-13(21)22)8-14(23)24/h9-10H,1-8H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)/p-2/t9-,10-/m1/s1

InChI key

FCKYPQBAHLOOJQ-NXEZZACHSA-L

Application


  • Polymer inclusion membranes as a new tool for Zn speciation: influence of the membrane composition in diffusional fluxes: This study explores the utilization of trans-1,2-diaminocyclohexane N,N,N′,N′-tetraacetic acid monohydrate (CDTA) in zinc speciation using polymer inclusion membranes (G Mostazo et al., 2017).

  • Stability, dissolution, and cytotoxicity of NaYF4-upconversion nanoparticles with different coatings: This research mentions the use of trans-1,2-diaminocyclohexane-N,N,N′,N′-tetracetic acid (CDTA) in the context of nanoparticle stabilization (V Bastos et al., 2022).

  • Amino acid complexes of zirconium in a carbon composite for the efficient removal of fluoride ions from water: Details the preparation using trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate in the development of a carbon composite for fluoride ion removal (E Gonzalez-Aguinaga et al., 2022).

  • Unusually Kinetically Inert Monocationic Neptunyl Complex with a Fluorescein-Modified 1, 10-Phenanthroline-2, 9-dicarboxylate Ligand: Specific Separation: Uses trans-1,2-diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate (CyDTA) as a masking agent in complexation studies (K Yamagata et al., 2023).

  • Selective extraction of polysaccharide affects the adsorption of proanthocyanidin by grape cell walls: This paper investigates the role of trans-1,2-diaminocyclohexane N,N,N′,N′-tetracetic acid monohydrate in the extraction process affecting the adsorption of proanthocyanidins (Y Ruiz-Garcia et al., 2014).

Analysis Note

Assay (complexometric, calc. on anhydrous substance): ≥ 99.0 %
Water (K. F.): 4.8 - 5.6 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service