Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.21007

Sigma-Aldrich

Biphenyl-2-ol

for synthesis

Synonym(s):

Biphenyl-2-ol, 2-Diphenylol, 2-Phenylphenol, 2-Hydroxydiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4-2-(OH)
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352104
EC Index Number:
201-993-5

vapor pressure

0.4 hPa ( 20 °C)

Quality Level

200
300

Assay

≥99.0% (GC)

form

powder

autoignition temp.

>520 °C

potency

>2000 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.4-9.5 % (v/v)

pH

7 (20 °C, 0.1 g/L in H2O)

bp

286 °C/1013 hPa

mp

56-58 °C

transition temp

flash point 138 °C

density

1.26 g/cm3 at 20 °C

bulk density

600 kg/m3

SMILES string

Oc1c(cccc1)c2ccccc2

InChI

1S/C12H10O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-9,13H

InChI key

LLEMOWNGBBNAJR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 56 °C
Melting range (upper value): ≤ 58 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(C)

124 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service