Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20969

Sigma-Aldrich

Perylene

for synthesis

Synonym(s):

Perylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
MDL number:
UNSPSC Code:
12352005
EC Index Number:
205-900-9
NACRES:
NA.22

vapor pressure

<0.001 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (HPLC)

form

crystals

mp

273-278 °C

storage temp.

2-30°C

SMILES string

c21c3c4c5c(c2cccc1ccc3)cccc5ccc4

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H

InChI key

CSHWQDPOILHKBI-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 273 °C
Melting range (upper value): ≤ 278 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service